Skip to Content
Merck
All Photos(1)

Key Documents

444605

Sigma-Aldrich

Melittin

A 26-residue polypeptide from bee venom that binds calmodulin in a Ca2+-dependent manner.

Synonym(s):

Melittin

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C131H229N39O31
CAS Number:
Molecular Weight:
2846.46
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77
Pricing and availability is not currently available.

Quality Level

description

Merck USA index - 14, 5824

Assay

>97% (HPLC)

form

lyophilized

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

solubility

water: 1 mg/mL

shipped in

ambient

storage temp.

−20°C

SMILES string

N1(C(CCC1)C(=O)NC(C)C(=O)NC(CC(C)C)C(=O)NC(C(CC)C)C(=O)NC(CO)C(=O)NC(Cc2c3c([nH]c2)cccc3)C(=O)NC(C(CC)C)C(=O)NC(CCCCN)C(=O)NC(CCCN\C(=N\[H])\N)C(=O)NC(CCCCN)C(=O)NC(CCCN\C(=N\[H])\N)C(=O)NC(CCC(=O)N)C(=O)NC(CCC(=O)N)C(=O)N)C(=O)C(NC(=O)CNC(=O)C(NC(=O)C(NC

InChI

1S/C131H229N39O31/c1-23-71(16)102(163-97(176)60-135)122(194)146-62-98(177)148-74(19)109(181)164-100(69(12)13)124(196)160-88(55-65(4)5)116(188)155-84(41-30-33-51-134)115(187)165-101(70(14)15)125(197)161-90(57-67(8)9)118(190)168-106(77(22)173)128(200)169-105(76(21)172)123(195)147-63-99(178)150-92(58-68(10)11)129(201)170-54-36-44-94(170)121(193)149-75(20)108(180)158-89(56-66(6)7)117(189)166-104(73(18)25-3)127(199)162-93(64-171)120(192)159-91(59-78-61-145-80-38-27-26-37-79(78)80)119(191)167-103(72(17)24-2)126(198)157-83(40-29-32-50-133)111(183)154-85(42-34-52-143-130(139)140)112(184)152-82(39-28-31-49-132)110(182)153-86(43-35-53-144-131(141)142)113(185)156-87(46-48-96(137)175)114(186)151-81(107(138)179)45-47-95(136)174/h26-27,37-38,61,65-77,81-94,100-106,145,171-173H,23-25,28-36,39-60,62-64,132-135H2,1-22H3,(H2,136,174)(H2,137,175)(H2,138,179)(H,146,194)(H,147,195)(H,148,177)(H,149,193)(H,150,178)(H,151,186)(H,152,184)(H,153,182)(H,154,183)(H,155,188)(H,156,185)(H,157,198)(H,158,180)(H,159,192)(H,160,196)(H,161,197)(H,162,199)(H,163,176)(H,164,181)(H,165,187)(H,166,189)(H,167,191)(H,168,190)(H,169,200)(H4,139,140,143)(H4,141,142,144)

InChI key

VDXZNPDIRNWWCW-UHFFFAOYSA-N

General description

A 26-residue polypeptide from bee venom that binds calmodulin in a Ca2+-dependent manner, activates phospholipase A2, and inhibits protein kinase C (IC50 = 5-7 µM) by binding to the catalytic domain in a Mg2+-ATP sensitive manner. A stimulator of Giα and G11α activity that is reported to inhibit adenylate cyclase activity in synaptic membranes. Has also been used for affinity purification of several Ca2+-binding proteins. Activates Giα-1 and G11α activities and inhibits Gs activity.

Biochem/physiol Actions

Cell permeable: no
Product does not compete with ATP.
Reversible: no
Target IC50: 5-7 µM 5-7 µM for activating phospholipase A2 and inhibits protein kinase C

Packaging

Yes

Warning

Toxicity: Standard Handling (A)

Sequence

H-Gly-Ile-Gly-Ala-Val-Leu-Lys-Val-Leu-Thr-Thr-Gly-Leu-Pro-Ala-Leu-Ile-Ser-Trp-Ile-Lys-Arg-Lys-Arg-Gln-Gln-NH₂

Other Notes

Fukushima, N., et al. 1998. Peptides19, 811.
Gravitt, K.R. 1994. Biochem. Pharmacol. 47, 425.
Baudier, J., et al. 1987. Biochemistry26, 2886.
Kincaid, R.L., and Coulson, C.C. 1985. Biochem. Biophys. Res. Commun.133, 256.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service