Skip to Content
Merck
All Photos(1)

Key Documents

444229

Sigma-Aldrich

MMP-8, Human Neutrophil

Synonym(s):

Matrix Metalloproteinase 8, Human Neutrophil Collagenase, Collagenase-2

Sign Into View Organizational & Contract Pricing


About This Item

Enzyme Commission number:
UNSPSC Code:
12352202
NACRES:
NA.77
Pricing and availability is not currently available.

Assay

≥90% (SDS-PAGE)

Quality Level

form

liquid

specific activity

≥40 mU/mg protein

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
avoid repeated freeze/thaw cycles

foreign activity

other MMP, none detected

shipped in

wet ice

storage temp.

−70°C

General description

Native pro-MMP-8 from stimulated human neutrophils. MMP-8 is an N-linked, complex glycoprotein that preferentially hydrolyzes Type I collagen versus Types II and III. Activated MMP-8 is inhibited by TIMP-1. Requires activation just prior to use. MW 58800/44000.

Packaging

Please refer to vial label for lot-specific concentration.

Warning

Toxicity: Standard Handling (A)

Other Notes

Deschamps, A. M., et al. 2005. Circulation111, 1166.
Mallya, S.K., et al. 1990. Biochemistry 29, 10628.
Marcy, A.I., et al. 1991. Biochemistry30, 6476.
Stricklin, et al. 1983. Biochemistry22, 61.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

  1. MMP-8 Human Neutrophil in which buffer it can be dissolve. For checking activity of MMP with collagen.

    1 answer
    1. This product is a 100ug/mL solution in 200 mM NaCl, 50 mM Tris-HCl, 5 mM CaCl?, 1 µM ZnCl?, 0.05% BRIJ® 35 Detergent, 0.05% NaN?, pH 7.0. See the link below for a general protocol. The researcher would have to optimize for use specifically with collagen.
      https://www.emdmillipore.com/US/en/product/MMP-8-Human-Neutrophil,EMD_BIO-444229#anchor_PDS

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service