Skip to Content
Merck
All Photos(1)

Key Documents

440202

Sigma-Aldrich

LY 294002

≥98% (HPLC), solid, PI3K inhibitor, Calbiochem®

Synonym(s):

LY 294002, 2-(4-Morpholinyl)-8-phenyl-4H-1-benzopyran-4-one, BRD2 Inhibitor IV, BRD3 Inhibitor III, BRD4 Inhibitor IV

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H17NO3
CAS Number:
Molecular Weight:
307.34
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77
Pricing and availability is not currently available.

Product Name

LY 294002, LY294002, CAS 154447-36-6, is a cell-permeable, potent, reversible, and specific inhibitor of PI 3-kinase ((IC₅₀ = 1.4 µM). Acts on the ATP-binding site.

Quality Level

Assay

≥98% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

off-white

solubility

DMSO: 20 mg/mL
ethanol: soluble

shipped in

ambient

storage temp.

−20°C

SMILES string

N4(CCOCC4)c1[o]c2c([c](c1)=O)cccc2c3ccccc3

InChI

1S/C19H17NO3/c21-17-13-18(20-9-11-22-12-10-20)23-19-15(7-4-8-16(17)19)14-5-2-1-3-6-14/h1-8,13H,9-12H2

InChI key

CZQHHVNHHHRRDU-UHFFFAOYSA-N

General description

A cell-permeable, potent, reversible and specific phosphatidylinositol 3-kinase inhibitor (IC50 = 1.4 µM) that acts on the ATP-binding site of the enzyme. Also inhibits nonhomologous DNA end-joining (NHEJ) activity of the 460 kDa phosphatidylinositol 3-like kinase DNA-PKcs, which is the catalytic subunit of DNA-activated protein kinase. Does not affect the activities of EGF receptor kinase, MAP kinase, PKC, Pl 4-kinase, S6 kinase, and c-Src even at 50 µM. Promotes the nuclear accumulation of active GSK-3b. Reported to suppress the proliferation of murine embryonic stem cells. Blocks the proliferation of cultured rabbit aortic smooth muscle cells without inducing apoptosis. Shown to bind to the same site as I-BET (Cat. No. 40101050 ~ 10 µM). A 10 mM (1 mg/325 µl) solution of LY 294002 (Cat. No. 440204) in DMSO is also available.
A cell-permeable, potent, reversible, and specific phosphatidylinositol 3-kinase (PI 3-kinase) inhibitor that acts on the ATP-binding site of the enzyme (IC50 = 1.4 µM). Also inhibits nonhomologous DNA end-joining (NHED) in the 460 kDa phosphatidylinositol 3-like kinase DNA-PKcs, which is the catalytic subunit of DNA-activated protein kinase. Does not affect the activity of EGF receptor kinase, MAP kinase, PKC, PI 4-kinase, S6 kinase, and c-src at 50 µM. Promotes the nuclear accumulation of active GSK-3b. Reported to suppress the proliferation of murine embryonic stem cells. Blocks the proliferation of cultured rabbit aortic smooth muscle cells without inducing apoptosis. Reported to inhibit histamine secretion in RBL-2H3 cells (IC50 = 5 µM).

Biochem/physiol Actions

Cell permeable: yes
Primary Target
Phosphatidylinositol 3-kinase
Product competes with ATP.
Reversible: yes
Target IC50: 1.4 µM against phosphatidylinositol 3-kinase

Warning

Toxicity: Standard Handling (A)

Preparation Note

Slight warming may be required to achieve complete solubilization.

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 6 months at -20°C.

Other Notes

Dittman, A., et al. 2013. ACS Chem. Biol.9, 495.
Bechard, M., and Dalton. S. 2009. Mol. Cell. Biol.29, 2092.
Lianguzova, M.S. et al. 2007. Cell Biol. Int.31, 330.
Baumann, P., and West, S.C. 1998. Proc. Natl. Acad. Sci. USA 95, 14066.
Cardone, M.H., et al. 1998. Science282, 1318.
Vlahos, C.J., et al. 1995. J. Immunol.154, 2413.
Yano, H., et al. 1995. Biochem. J.312, 145.
Vlahos, C.J., et al. 1994. J. Biol. Chem.269, 5241.



Selected Citations
Lee, J., et al. 2009. Cell Stem Cell5, 76.


Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

487.4 °F

Flash Point(C)

253.0 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chengping Li et al.
International journal of oncology, 48(6), 2639-2646 (2016-04-02)
TRIP-Br1 oncoprotein is known to be involved in many vital cellular functions. In this study, we examined the role of TRIP-Br1 in hypoxia-induced cell death. Exposure to the overcrowded and CoCl2-induced hypoxic conditions increased TRIP-Br1 expression at the protein level
Shu Qi et al.
Cancer letters, 483, 98-113 (2020-03-29)
Liver metastases remain a major cause of death from gastrointestinal tract cancers and other malignancies, such as breast and lung carcinomas. Understanding the underlying biology is essential for the design of effective therapies. We previously identified the chemokine CCL7 and
Aadhya Tiwari et al.
Cancers, 12(10) (2020-10-03)
The multifunctional protein Y-box binding protein-1 (YB-1) regulates all the so far described cancer hallmarks including cell proliferation and survival. The MAPK/ERK and PI3K/Akt pathways are also the major pathways involved in cell growth, proliferation, and survival, and are the
Kelly H Forest et al.
Frontiers in molecular neuroscience, 14, 576038-576038 (2021-04-30)
Alzheimer's disease (AD) is the most common cause of dementia in the aging population. Evidence implicates elevated soluble oligomeric Aβ as one of the primary triggers during the prodromic phase leading to AD, effected largely via hyperphosphorylation of the microtubule-associated
Milica Bozic et al.
Nature communications, 11(1), 1943-1943 (2020-04-25)
Kidney fibrosis is a highly deleterious process and a final manifestation of chronic kidney disease. Alpha-(α)-synuclein (SNCA) is an actin-binding neuronal protein with various functions within the brain; however, its role in other tissues is unknown. Here, we describe the

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service