Skip to Content
Merck
All Photos(1)

Key Documents

400076-M

Millipore

Hypericin

Polycyclic dione that has anti-retroviral activity.

Synonym(s):

Hypericin, Casein Kinase II Inhibitor XI

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C30H16O8
CAS Number:
Molecular Weight:
504.44
MDL number:
UNSPSC Code:
12352200
Pricing and availability is not currently available.

description

Merck USA index - 14, 4863

Quality Level

Assay

≥98% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

black

solubility

ethanol: 1 mg/mL
methanol: 1 mg/mL
DMSO: 10 mg/mL

storage temp.

2-8°C

SMILES string

Oc1c2c3c4c5c6c2c(cc(c6C(=O)c7c5c(c8c4c(c(cc8C)O)C(=O)c3c(c1)O)c(cc7O)C)O)O

InChI

1S/C30H16O8/c1-7-3-9(31)19-23-15(7)16-8(2)4-10(32)20-24(16)28-26-18(12(34)6-14(36)22(26)30(20)38)17-11(33)5-13(35)21(29(19)37)25(17)27(23)28/h3-6,31-36H,1-2H3

InChI key

BTXNYTINYBABQR-UHFFFAOYSA-N

General description

Polycyclic dione that has anti-retroviral activity. Inhibits protein kinase C (IC50 = 3.3 µM). Also known to inhibit the protein tyrosine kinase activities of the insulin receptor (IC50 = 20-29 nM), the epidermal growth factor receptor (IC50 = 35 nM), casein kinase II (IC50 = 6 nM), and MAP kinase (IC50 = 4 nM). Useful probe for PKC due to its bright red fluorescence emission and photostability. Also see Pseudohypericin (Cat. No. 539590).

Biochem/physiol Actions

Primary Target
PKC
Target IC50: 3.3 µM, 6 nM, 4 nM, 20-29 nM , 35 nM, against protein kinase C, casein kinase II, MAP kinase, the protein tyrosine activities of the insulin receptor, and the epidermal growth factor receptor, respectively

Warning

Toxicity: Harmful (C)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Zhang, W., et al. 1997. Cancer Lett.120, 31.
Agostinis, P., et al. 1995. Biochem. Pharmacol.49, 1615.
Frew, T., et al. 1994. Anticancer Res.14, 2425.
Barnard, D.L., et al. 1992. Antiviral Res.17, 63.
Takahashi, I., et al. 1989. Biochem. Biophys. Res. Commun.165, 1207.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service