Skip to Content
Merck
All Photos(1)

Key Documents

233501-M

Sigma-Aldrich

Coagulation Factor XIII, Human Plasma

Coagulation Factor XIII, Human Plasma, CAS 9013-56-3, is a native tetrameric form composed of two identical α- and two β-subunits. Thrombin converts the α2β2 tetramer to a modified α2ʹβ2 tetramer.

Synonym(s):

Coagulation Factor XIII, Human Plasma, Fibrin-stabilizing Factor

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352202
Pricing and availability is not currently available.

Quality Level

description

Merck USA index - 14, 3926

Assay

≥95% (SDS-PAGE)

form

liquid

specific activity

≥40 units/mg protein (clotting assay)

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

storage temp.

−20°C

General description

Native coagulation factor XIII from human plasma. Plasma Factor XIII is a tetramer (α2β2) composed of two identical α-subunits and two identical β-subunits. After thrombin cleavage converts the α2β2 tetramer to a modified α2′β2 tetramer, Ca2+ promotes both the dissociation of the β2 dimer and the activation of the α2′ dimer to its catalytically active form. This activated Factor XIII (Factor XIIIa) strengthens the fibrin clot by catalyzing the formation of glutamyl-lysine bonds between fibrin monomers.

Warning

Toxicity: Standard Handling (A)

Unit Definition

Clotting activity is determined by comparison to a standard curve produced using normal pooled human plasma. One unit is defined as the activity in 1 ml of normal, pooled plasma.

Physical form

In 500 µM EDTA, 50% glycerol.

Preparation Note

Prepared from plasma that has been shown by certified tests to be negative for HBsAg and for antibodies to HIV and HCV.

Reconstitution

Following initial thaw, aliquot and freeze (-20°C).

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service