18-161
Protein G, HRP conjugate
500 µg Protein G, HRP conjugate for Dot Blot, ELISA, Western Blotting & IHC.
Synonym(s):
Protein G HRP
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41105332
eCl@ss:
32160405
NACRES:
NB.22
Recommended Products
form
solid
manufacturer/tradename
Upstate®
technique(s)
ELISA: suitable
dot blot: suitable
immunohistochemistry: suitable
western blot: suitable
shipped in
wet ice
Application
Suitable for immunoblotting (western or dot blot), ELISA, immunoperoxidase electron microscopy, and immunohistochemistry.
Quality
Suitable for immunoblotting (western or dot blot), ELISA, immunoperoxidase electron microscopy, and immunohistochemistry
Physical form
chromatographically pure Protein G conjugated to horseradish peroxidase lyophilized from 0.02 M Potassium Phosphate, 0.15 M NaCl, pH 7.2, 10 mg/ml BSA.
Storage and Stability
Lyophilized: Stable for 2 years at -20°C from date of shipment. Rehydrated: Stable for 6 months at -20°C
Legal Information
UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany
Disclaimer
Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Natalia Sanchez de Groot et al.
Molecular systems biology, 15(4), e8075-e8075 (2019-04-10)
Phase separation of soluble proteins into insoluble deposits is associated with numerous diseases. However, protein deposits can also function as membrane-less compartments for many cellular processes. What are the fitness costs and benefits of forming such deposits in different conditions?
Ryan J H West et al.
Human molecular genetics, 27(8), 1382-1395 (2018-02-13)
Frontotemporal dementia (FTD) is one of the most prevalent forms of early-onset dementia. However, the pathological mechanisms driving neuronal atrophy in FTD remain poorly understood. Here we identify a conserved role for the novel pro-apoptotic protein plenty of SH3s (POSH)/SH3
Guillaume Lefebvre et al.
Pharmaceutical research, 38(12), 2157-2166 (2021-12-15)
Monoclonal antibodies are in contact with many different materials throughout their life cycle from production to patient administration. Plastic surfaces are commonly found in single use bags, syringes, perfusion bags and tubing and their hydrophobic nature makes them particularly prone
First-in-human autologous implantation of genetically modified adipocytes expressing LCAT for the treatment of familial LCAT deficiency.
Aso, et al.
Heliyon, 8, e11271-e11271 (2022)
Aida Barreiro-Alonso et al.
Cancers, 13(18) (2021-09-29)
This study reports the HMGB1 interactomes in prostate and ovary cancer cells lines. Affinity purification coupled to mass spectrometry confirmed that the HMGB1 nuclear interactome is involved in HMGB1 known functions such as maintenance of chromatin stability and regulation of
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service