Skip to Content
Merck
All Photos(2)

Key Documents

1.46406

Millipore

Fluid Thioglycollate Medium

bottle capacity 100 mL, bottle filling volume 100 mL, closure type, Red flip cap with septum, pack of 10 bottles

Synonym(s):

Fluid Thioglycollate Medium acc EP+USP, Fluid Thioglycollate Medium, FTM, Thioglycollate Broth

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106200
NACRES:
NA.74
Pricing and availability is not currently available.

Agency

EP 2.6.1
JP 4.06
USP 71

Quality Level

sterility

sterile; autoclaved

form

liquid (ready-to-use)

shelf life

12 mo.

feature

closure type Red flip cap with septum
ready-to-use

packaging

pack of 10 bottles

bottle capacity

100 mL

bottle filling volume

100 mL

capacity

100 mL

pH

7.1

application(s)

cosmetics
food and beverages
pharmaceutical
sterility testing

compatibility

for use with Steritest® Symbio FLEX Pump Kit, 2 media (SYMBFLE01)
for use with Steritest® Symbio ISL Pump Kit, 2 media (SYMBISL01)
for use with Steritest® Symbio LFH Pump Kit (SYMBLFH01)

storage temp.

2-25°C

suitability

aerobic bacteria
anaerobic bacteria

Looking for similar products? Visit Product Comparison Guide

General description

Fluid Thioglycolate Medium (FTM) is a universal complex medium. Thioglycolate and L-Cystine in the medium reduce the redox potential to create an anaerobic atmosphere. Additionally, they inactivate mercury and other heavy metal compounds. Agar further reduces the rapid diffusion of oxygen through the medium but may lead to slight turbidity in larger volumes (filled bottles). Resazurin indicates the reduction potential of the medium, and a color change to pink indicates an increased concentration of oxygen.

Application

Fluid Thioglycolate Medium is suitable for the isolation and cultivation of fastidious anaerobic as well as aerobic microorganisms. It is used for sterility control of pharmaceutical products.

Features and Benefits

Our ready to use media provide the highest level of quality and testing confidence. They have been formulated and tested to meet the pharmacopoeia requirements. Sterility testing media and rinse solutions are manufactured in an ISO 9001, environmentally controlled production center. Each lot undergoes a stringent quality control (QC) procedure, including pH, sterility and growth promotion tests. Our manufacturing approach ensures the highest level of clarity for our media and rinsing fluids, therefore improving accuracy and significantly reducing the risk of incorrect interpretation and false results.

Legal Information

STERITEST is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service