1.46224
Clear Fluid Thioglycollate Medium
tube of, ready-to-use, tube volume 15 mL , filling volume
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
Recommended Products
Agency
EP
JP
USP
Quality Level
sterility
sterile; autoclaved
form
solution
feature
closure type screw cap with septum
ready-to-use
packaging
single packed of
tube of
tube capacity
25 mL
tube volume
15 mL , filling volume
application(s)
pharmaceutical
sterility testing
General description
FTM 15 ml in 25 ml tube.
Fluid Thioglycollate Medium is a universal complex medium. Thioglycollate and L-Cystine reduce the redox potential of the culture medium and create an anaerobic atmosphere, along with inactivating mercury and other heavy metal compounds. The presence of agar reduces a rapid diffusion of oxygen through the medium but may lead to slight turbidity in larger volumes (filled bottles). Resazurin indicates the reduction potential of the medium. An increased concentration of oxygen is indicated by a color change to pink.
Fluid Thioglycollate Medium is a universal complex medium. Thioglycollate and L-Cystine reduce the redox potential of the culture medium and create an anaerobic atmosphere, along with inactivating mercury and other heavy metal compounds. The presence of agar reduces a rapid diffusion of oxygen through the medium but may lead to slight turbidity in larger volumes (filled bottles). Resazurin indicates the reduction potential of the medium. An increased concentration of oxygen is indicated by a color change to pink.
Application
Fluid Thioglycollate Medium is used for the isolation and cultivation of fastidious anaerobic and aerobic microorganisms. It is suitable for sterility control of pharmaceutical products.
related product
Storage Class Code
10 - Combustible liquids
WGK
WGK 2
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service