Skip to Content
Merck
All Photos(2)

Key Documents

1.46005

Millipore

Sabouraud Dextrose Agar

Sodium thiosulphate, Tween®, Histidine, Lecithin, lid, ICR: non lockable 30 mL, plate diam. 90 mm, sterile; irradiated, triple packed of 20 or 120 plates, suitable for air monitoring

Synonym(s):

SDA, SDA ICR plate, SDA with Neutralizers

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106200
NACRES:
NA.74

material

polystyrene plate

Quality Level

Agency

EP
USP

sterility

sterile; irradiated

form

plate (ready-to-use)

feature

lid ICR: non lockable 30 mL
ready-to-use

composition

Histidine
Lecithin
Sodium thiosulphate
Tween®

packaging

triple packed of 20 or 120 plates

plate diam.

90 mm

pH

5.6 ( in H2O)

application(s)

air monitoring
pharmaceutical

compatibility

for use with MAS-100®

storage temp.

2-25°C

suitability

molds
yeasts

Looking for similar products? Visit Product Comparison Guide

General description

Sabouraud Dextrose Agar + LTHTh - ICR in 90 mm settle plates (non- lockable) is designed for the determination of the total count of yeasts and molds in air via active or passive air monitoring as well as fingerprints of personnel in critical clean room classes or isolators. Sabouraud Dextrose Medium contains digests of animal tissues (peptones) which provide a nutritious source of amino acids and nitrogenous compounds for the growth of fungi and yeasts. Dextrose is added as the energy and carbon source. The low pH favours fungal growth. Lecithin, polysorbate 80 (Tween®), histidine and sodium thiosulfate are neutralizers to inactivate residues of disinfectants. To differentiate Tryptic Soy Agar (TSA) from Sabouraud Dextrose agar (SDA), SDA ICR media are filled in pink colored dish.

Application

Sabouraud Dextrose Agar (SDA) is a complex medium for cultivation and isolation of yeasts and molds. The media is used for environmental monitoring by determining the yeast and mold count in air through active or passive air monitoring as well as fingerprints of personnel in isolators and clean rooms.

Legal Information

MAS-100 is a registered trademark of MBV AG
TWEEN is a registered trademark of Croda International PLC

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service