14-530-M
B-Raf (δ1-415) Protein, active, 10 µg
Active, N-terminal GST-tagged recombinant human B-Raf residues 416-end, for use in Kinase Assays.
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
biological source
human
Quality Level
recombinant
expressed in baculovirus infected Sf21 cells
mol wt
Mw 67.2 kDa
manufacturer/tradename
Upstate®
technique(s)
activity assay: suitable (kinase)
NCBI accession no.
UniProt accession no.
General description
N-terminal GST-tagged recombinant human B-Raf residues 416-end
Product Source: Expressed by baculovirus in Sf21 insect cells
Application
Research Category
Metabolism
Inflammation & Immunology
Metabolism
Inflammation & Immunology
Research Sub Category
Obesity
Metabolic Disorders
Osteoporosis
Arthritis
Obesity
Metabolic Disorders
Osteoporosis
Arthritis
Biochem/physiol Actions
Protein Target: B-Raf
Target Sub-Family: TKL
Packaging
Also available in 250 µg size --call for pricing and availability and reference catalog number 14-530M when ordering the 250 µg size.
Quality
routinely evaluated by phosphorylation of MBP substrate in vitro
Physical form
Glutathione agarose affinity chromatography
Storage and Stability
6 months at -70°C
Other Notes
For Specific Activity data, refer to the Certificate of Analysis for individual lots of this enzyme.
Legal Information
UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany
Disclaimer
Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Cell, 116(6), 764-766 (2004-03-24)
A study by Wan et al. in this issue of Cell demonstrates that the majority of oncogenic mutations in the B-Raf protein kinase result in increased catalytic activity, through disruption of the autoinhibited state of the kinase domain. Surprisingly, several
Cancer metastasis reviews, 24(1), 165-183 (2005-03-24)
Melanoma is a form of skin cancer that has a poor prognosis and which is on the rise in Western populations. If detected early, it is easily treated by surgical excision. However, once melanoma metastasises it is notoriously resistant to
Cancer cell, 6(4), 313-319 (2004-10-19)
RAF proteins are components of a conserved signaling pathway that regulates cellular responses to extracellular signals. Despite over 20 years of research into the regulation and function of the RAF proteins, it was only realized recently that the B-RAF isoform
Biochimica et biophysica acta, 1653(1), 25-40 (2003-06-05)
A recent report has shown that activating mutations in the BRAF gene are present in a large percentage of human malignant melanomas and in a proportion of colon cancers. The vast majority of these mutations represent a single nucleotide change
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service