Skip to Content
Merck
All Photos(1)

Key Documents

1.17179

Supelco

Carbon Dioxide Test Kit, titrimetric

titrimetric, for use with MCOLORTEST®

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116116
NACRES:
NB.21
Pricing and availability is not currently available.

Product Name

Carbon Dioxide Test, titrimetric, for use with MCOLORTEST®

specific analyte(s)

carbon dioxide

Quality Level

compatibility

for use with MCOLORTEST®

detection method

titrimetric

storage temp.

15-25°C

General description

This MQuant® Carbon Dioxide test (formally MColortest®) is suitable for the titrimetric determination using a convenient dropping bottle.

Method
The sample is titrated with sodium hydroxide solution against phenolphthalein until its color changes to pink to red (at pH 8.2). The carbon dioxide content is determined from the consumption of titration solution.

The test can be used in
  • Groundwater and surface water, seawater
  • Drinking water
  • Industrial water
  • Boiler water
  • Wastewater
method: titrimetric MColortest®

Legal Information

MCOLORTEST is a registered trademark of Merck KGaA, Darmstadt, Germany
MQUANT is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Statements

Hazard Classifications

Carc. 1B - Eye Irrit. 2 - Flam. Liq. 2 - Muta. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

71.6 °F

Flash Point(C)

22 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service