1.15525
RINGER tablets
for the preparation of RINGER′S solution
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Quality Level
sterility
non-sterile
form
tablet
technique(s)
microbiological culture: suitable
pH
7 (20 °C, 2.5 g/L in H2O)
application(s)
food and beverages
microbiology
storage temp.
15-25°C
General description
RINGER tablets are used for the preparation of the RINGER′S solution. Ringer Salt Solution is isotonic with bacteria and thus prevents them from being subjected to osmotic stress when they are removed from their customary environment. It is physiologically superior to physiological saline for sensitive organisms and preserves the cells in their original condition. The salts in the medium balance the osmotic equilibrium of the medium, thereby protecting the organisms from osmotic stress caused due to change in the environment.
Application
RINGER tablets are used for the preparation of the RINGER′S solution, which is used as an isotonic diluent for food, milk, and dairy products during the microbiological examination. RINGER solution is utilized like a diluent for preparing suspensions in bacteriological studies, especially in the examination of milk and dairy products.
Analysis Note
Appearance of solution (after autoclave): clear
pH-value (after autoclave): 6.8 - 7.2
Weight (min): ≥ 1.47 g
Weight (max): ≤ 1.79 g
Suitability (microbiological): passes test
pH-value (after autoclave): 6.8 - 7.2
Weight (min): ≥ 1.47 g
Weight (max): ≤ 1.79 g
Suitability (microbiological): passes test
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2
Storage Class Code
13 - Non Combustible Solids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service