Skip to Content
Merck
All Photos(1)

Key Documents

1.12144

Supelco

Disulfine blue VN 150 (C.I.42045)

for surfactant tests

Synonym(s):

Disulfine blue VN 150 (C.I.42045)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C27H31N2NaO6S2
CAS Number:
Molecular Weight:
566.66
MDL number:
UNSPSC Code:
12352107
EC Index Number:
204-934-1
NACRES:
NA.21
Pricing and availability is not currently available.

Quality Level

composition

dye content, ≥40% spectrophotometric assay

loss

≤7% loss on drying, 110°C

pH

6.9 (20 °C, 10 g/L in H2O)

mp

290 °C (decomposition)

solubility

210 g/L

bulk density

600 kg/m3

λmax

637-640 nm in water

storage temp.

2-30°C

SMILES string

[Na+].[S](=O)(=O)([O-])c1c(ccc(c1)[S](=O)(=O)[O-])C(=C3C=CC(=[N+](CC)CC)C=C3)c2ccc(cc2)N(CC)CC

InChI

1S/C27H32N2O6S2.Na/c1-5-28(6-2)22-13-9-20(10-14-22)27(21-11-15-23(16-12-21)29(7-3)8-4)25-18-17-24(36(30,31)32)19-26(25)37(33,34)35;/h9-19H,5-8H2,1-4H3,(H-,30,31,32,33,34,35);/q;+1/p-1

InChI key

SJEYSFABYSGQBG-UHFFFAOYSA-M

Analysis Note

Dye content (spectrophotometrically): ≥ 40 %
Identity (UV/VIS-Spectrum): passes test
Absorption maximum λmax. (water): 637 - 640 nm
Spec. Absorptivity A 1%/1cm (λmax; 0.005 g/l; water): ≥ 800
TLC-Test: passes test
Loss on drying (110 °C): ≤ 7 %
Suitability for surfactant tests: passes test

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service