1.09991
Zinc sulfate solution
c(ZnSO₄) = 0.1 mol/l (0.1 N), concentrate for preparing 1000 ml volumetric solution for titration, Titrisol®
Synonym(s):
ZnSO4 solution
About This Item
Recommended Products
Product Name
Zinc sulfate solution, for 1000 ml, c(ZnSO₄) = 0.1 mol/l (0.1 N) Titrisol®
grade
reagent grade
Quality Level
Agency
reag. Ph. Eur.
product line
Titrisol®
form
liquid
reaction suitability
reaction type: Redox Reactions
concentration
0.1 M
technique(s)
titration: suitable
pH
2.2 (20 °C in H2O)
density
1.16 g/cm3 at 20 °C
storage temp.
15-25°C
Application
- Enhanced photocatalytic dye degradation using surface-modified tungstophosphoric acid-iron magnetic nanocatalyst.: This paper presents the development and application of a surface-modified tungstophosphoric acid-iron magnetic nanocatalyst for the enhanced photocatalytic degradation of dyes. The study highlights the catalyst′s high efficiency and potential for environmental remediation and wastewater treatment (Kaplaner et al., 2024).
Legal Information
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Aquatic Chronic 2 - Eye Dam. 1 - Met. Corr. 1
Storage Class Code
8B - Non-combustible, corrosive hazardous materials
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service