Skip to Content
Merck
All Photos(1)

Key Documents

1.09989

Supelco

Nickel standard

1000 mg Ni, (NiCl₂ in H₂O) Titrisol®

Synonym(s):

Nickel standard for aqueous matrices

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.24
Pricing and availability is not currently available.

grade

analytical standard

Quality Level

Agency

suitable for EPA 200.7
suitable for EPA 200.8

product line

Certipur®

concentration

1000 mg Ni in H2O

technique(s)

AAS: suitable
ICP: suitable

pH

4.4 (20 °C in H2O)

density

1.04 g/cm3 at 20 °C

application(s)

industrial qc
pharmaceutical

format

single component solution

storage temp.

15-25°C

Application

Thenickel standard can be used as a calibration standard for the determination ofnickel by Flame Atomic Absorption Spectrometry (FAAS) following its separationfrom edible oils. [1]

Features and Benefits

  • Titrisol® offers a wide range of concentrated single-element standards.
  • Dilute the concentrated solution to achieve the desired standard concentration.
  • Directly traceable to standard reference materials (SRM) from the National Institute of Standards and Technology (NIST).
  • Supplied in easy-to-handle plastic ampoules.
  • Accompanied with a batch-specific certificate of analysis (COA) containing information on the NIST batch and minimum shelf life.
  • Provides reliable and reproducible results.

Analysis Note

content (Ni): 1000 mg/Ampoule
Determination method: Complexometric titration.
(traceable of NIST - SRM 682)
Accurcy of the method: +/- 2 mg

Legal Information

CERTIPUR is a registered trademark of Merck KGaA, Darmstadt, Germany
Titrisol is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Health hazardEnvironment

Signal Word

Danger

Hazard Classifications

Aquatic Chronic 2 - Carc. 1A Inhalation - Muta. 2 - Repr. 1B - Resp. Sens. 1 - Skin Sens. 1 - STOT RE 1 - STOT RE 2 Inhalation

Target Organs

Lungs

Storage Class Code

6.1D - Non-combustible, acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Related Content

This page is intended to make it easier to find the consumables you need based on the analytical method you’re using. Methods included on this page come from the EPA, Standard Methods and ASTM.

This page is intended to make it easier to find the consumables you need based on the analytical method you’re using. Methods included on this page come from the EPA, Standard Methods and ASTM.

This page is intended to make it easier to find the consumables you need based on the analytical method you’re using. Methods included on this page come from the EPA, Standard Methods and ASTM.

This page is intended to make it easier to find the consumables you need based on the analytical method you’re using. Methods included on this page come from the EPA, Standard Methods and ASTM.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service