Skip to Content
Merck
All Photos(1)

Key Documents

108976

Sigma-Aldrich

Leupeptin, Hemisulfate, Synthetic

Synthetic version of the microbial-derived inhibitor Leupeptin against trypsin-like and cysteine protease activities.

Synonym(s):

Leupeptin, Hemisulfate, Synthetic, Ac-LLR-CHO, 1/2H₂SO₄

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H38N6O4 · 0.5H2SO4
CAS Number:
Molecular Weight:
475.59
MDL number:
UNSPSC Code:
12352202
NACRES:
NA.21
Pricing and availability is not currently available.

description

Merck USA index - 14, 5456.

Quality Level

Assay

≥95% (sum of isomers, HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated (hygroscopic)

color

off-white

solubility

water: 50 mg/mL

shipped in

ambient

storage temp.

−20°C

SMILES string

[N+H](=C(N)N)CCC[C@H](NC(=O)[C@@H](NC(=O)[C@@H](NC(=O)C)CC(C)C)CC(C)C)C=O

InChI

1S/C20H38N6O4/c1-12(2)9-16(24-14(5)28)19(30)26-17(10-13(3)4)18(29)25-15(11-27)7-6-8-23-20(21)22/h11-13,15-17H,6-10H2,1-5H3,(H,24,28)(H,25,29)(H,26,30)(H4,21,22,23)/p+1/t15-,16-,17-/m0/s1

InChI key

GDBQQVLCIARPGH-ULQDDVLXSA-O

General description

Synthetic version of the microbial-derived inhibitor Leupeptin (Cat. No. 108975) against trypsin-like and cysteine protease activities.

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Sequence

Ac-Leu-Leu-arginal, hemisulfate

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Gemma Gou et al.
Journal of neurochemistry, 154(6), 618-634 (2020-02-19)
The SynGAP protein is a major regulator of synapse biology and neural circuit function. Genetic variants linked to epilepsy and intellectual disability disrupt synaptic function and neural excitability. SynGAP has been involved in multiple signaling pathways and can regulate small
S Joseph Endicott et al.
The Journal of cell biology, 219(12) (2020-10-14)
Chaperone-mediated autophagy (CMA) is the most selective form of lysosomal proteolysis, where individual peptides, recognized by a consensus motif, are translocated directly across the lysosomal membrane. CMA regulates the abundance of many disease-related proteins, with causative roles in neoplasia, neurodegeneration
Pietri Puustinen et al.
Autophagy, 16(10), 1871-1888 (2020-01-28)
Macroautophagy/autophagy is a central component of the cytoprotective cellular stress response. To enlighten stress-induced autophagy signaling, we screened a human kinome siRNA library for regulators of autophagic flux in MCF7 human breast carcinoma cells and identified the catalytic subunit of

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service