Skip to Content
Merck
All Photos(1)

Key Documents

1.08238

Millipore

Glycerol triacetate

for biochemistry

Synonym(s):

Glycerol triacetate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H14O6
CAS Number:
Molecular Weight:
218.20
MDL number:
UNSPSC Code:
41141833
EC Index Number:
203-051-9
NACRES:
NA.79

vapor pressure

<0.1 hPa ( 20 °C)

Quality Level

form

liquid

autoignition temp.

430 °C

potency

3000 mg/kg LD50, oral (Rat)
>5000 mg/kg LD50, skin (Rabbit)

expl. lim.

1.1-7.7 % (v/v)

pH

5.0-6.0 (20 °C, 50 g/L in H2O)

bp

258 °C/1013 hPa

mp

-78 °C

transition temp

flash point 138 °C

solubility

64 g/L

density

1.16 g/cm3 at 20 °C

storage temp.

15-25°C

InChI

1S/C9H14O6/c1-6(10)13-4-9(15-8(3)12)5-14-7(2)11/h9H,4-5H2,1-3H3

InChI key

URAYPUMNDPQOKB-UHFFFAOYSA-N

Analysis Note

Purity (GC): ≥ 99 %
Identity (IR-spectrum): passes test
Density (d 20/4): 1.157 - 1.159
Water (according to Karl Fischer): ≤ 0.2 %
Suitability as enzyme substrate (tested with Lipase): passes test

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

298.4 °F - closed cup

Flash Point(C)

148 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service