Skip to Content
Merck
All Photos(1)

Key Documents

1.07826

Supelco

Tin

foil about 0.04 mm thick

Synonym(s):

Tin

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Sn
CAS Number:
Molecular Weight:
118.71
MDL number:
UNSPSC Code:
12141745
EC Index Number:
231-141-8
NACRES:
NA.21
form:
solid

Quality Level

form

solid

bp

2260 °C/1013 hPa

mp

232 °C

density

7.3 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

[Sn]

InChI

1S/Sn

InChI key

ATJFFYVFTNAWJD-UHFFFAOYSA-N

Related Categories

Analysis Note

Identity (Sn): positive

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service