Skip to Content
Merck
All Photos(1)

Key Documents

1.06309

Supelco

di-Sodium tetraborate

anhydrous 99.99 Suprapur®

Synonym(s):

di-Sodium tetraborate, Borax

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
B4Na2O7
CAS Number:
Molecular Weight:
201.22
MDL number:
UNSPSC Code:
12352302
EC Index Number:
215-540-4
NACRES:
NB.21
Assay:
99.99%
grade:
for inorganic trace analysis
solubility:
25.6 g/L
Pricing and availability is not currently available.

grade

for inorganic trace analysis

Quality Level

vapor pressure

7.3 hPa ( 1200 °C)

Assay

99.99%

potency

>2000 mg/kg LD50, skin (Rabbit)

pH

9.2 (20 °C, 25 g/L in H2O)

mp

742 °C

solubility

25.6 g/L

density

2.37 g/cm3 at 20 °C

bulk density

700 kg/m3

storage temp.

2-30°C

SMILES string

[Na+].[Na+].[B+3].[B+3].[B+3].[B+3].[O-2].[O-2].[O-2].[O-2].[O-2].[O-2].[O-2].O.O.O.O.O.O.O

InChI

1S/4B.2Na.7H2O.7O/h;;;;;;7*1H2;;;;;;;/q4*+3;2*+1;;;;;;;;7*-2

InChI key

HJIQVKXMQOLFFS-UHFFFAOYSA-N

Analysis Note

Purity (metallic): ≥ 99.99 %
Assay (acidimetric): ≥ 99 %
Chloride (Cl): ≤ 5 ppm
Phosphate (PO₄): ≤ 5 ppm
Silicate (SiO₂): ≤ 5 ppm
Sulfate (SO₄): ≤ 50 ppm
Ba (Barium): ≤ 10.0 ppm
Ca (Calcium): ≤ 5.0 ppm
Cd (Cadmium): ≤ 0.050 ppm
Ce (Cerium): ≤ 0.05 ppm
Co (Cobalt): ≤ 0.100 ppm
Cr (Chromium): ≤ 0.05 ppm
Cu (Copper): ≤ 0.10 ppm
Eu (Europium): ≤ 0.05 ppm
Fe (Iron): ≤ 0.10 ppm
K (Potassium): ≤ 50 ppm
La (Lanthanum): ≤ 0.05 ppm
Li (Lithium): ≤ 5.0 ppm
Mg (Magnesium): ≤ 0.10 ppm
Mn (Manganese): ≤ 0.050 ppm
Ni (Nickel): ≤ 0.05 ppm
Pb (Lead): ≤ 0.05 ppm
Rb (Rubidium): ≤ 5 ppm
Sc (Scandium): ≤ 0.05 ppm
Sm (Samarium): ≤ 0.05 ppm
Sr (Strontium): ≤ 5.00 ppm
Tl (Thallium): ≤ 0.05 ppm
Y (Yttrium): ≤ 0.05 ppm
Yb (Ytterbium): ≤ 0.050 ppm
Zn (Zinc): ≤ 0.05 ppm

Legal Information

SUPRAPUR is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation markHealth hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Repr. 1B

Storage Class Code

6.1D - Non-combustible, acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service