Skip to Content
Merck

1.05032

SAFC

Potassium hydroxide

pellets EMPROVE® ESSENTIAL Ph Eur,BP,JP,NF,FCC,E 525

Pharma Manufacturing

Synonym(s):

Potash caustic, Caustic potash

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
KOH
CAS Number:
Molecular Weight:
56.11
MDL number:
UNSPSC Code:
12352305
E Number:
E525
EC Index Number:
215-181-3

Agency

BP
JP
NF
Ph. Eur.

Quality Level

reg. compliance

FCC

vapor pressure

1 mmHg ( 719 °C)

product line

EMPROVE® ESSENTIAL

form

solid

potency

333 mg/kg LD50, oral (Rat)

pH

14 (20 °C, 56 g/L in H2O)

mp

361 °C (lit.)

application(s)

liquid formulation
pharmaceutical

storage temp.

15-25°C

SMILES string

[OH-].[K+]

InChI

1S/K.H2O/h;1H2/q+1;/p-1

InChI key

KWYUFKZDYYNOTN-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

We offer a range of high quality critical raw materials, solvents and reagents plus the documentation and expertise to meet stringent regulatory requirements. In addition to our comprehensive portfolio, we also offer custom manufacturing services to meet your exact needs. As part of our EMPROVE® Program, our raw materials are offered with extensive documentation facilitating compliance of your pharma and biopharma product, full supply chain transparency and risk mitigation. Our SAFC® portfolio of high-quality products for biopharmaceutical and pharmaceutical formulation and production withstands strict quality control procedures and is produced according to applicable cGMP guidelines.

Application

Potassium hydroxide pellets are used in pH-adjustment for various cremes and (baby-)food. Potassium hydroxide pellets Emprove® Essential is manufactured according to GMP guidelines and is suitable for pharmaceutical and biopharmaceutical processes.

Legal Information

Emprove is a registered trademark of Merck KGaA, Darmstadt, Germany
SAFC is a registered trademark of Merck KGaA, Darmstadt, Germany

also commonly purchased with this product

related product

Product No.
Description
Pricing

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1A

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service