Skip to Content
Merck
All Photos(1)

Key Documents

1.04447

Supelco

ICP Multi element standard solution XXIV certified reference material

(15 elements in diluted nitric acid) tuning solution 700 ES, Certipur®

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200

Quality Level

technique(s)

ICP: suitable

matrix

(in diluted nitric acid)

pH

1 (20 °C in water)

density

1.005 g/cm3 at 20 °C

application(s)

environmental
food and beverages
personal care
(industry & testing)

format

multi-component solution

storage temp.

15-25°C

General description

This certified reference material (CRM) is produced in accordance with ISO 17034 and characterized in accordance with  ISO/IEC 17025. This CRM is traceable to SI unit kg and measured against primary material from an NMI, e.g. NIST.   

Please visit  ISO certificates and Site Quality Self-Assessments to access the current certificates of accreditation.   

Download your certificate at  https://www.sigmaaldrich.com/US/en/documents-search?tab=coa  to view certified values, including uncertainty and expiry date.

Application

Multielement standard solution XXIV can be used as a calibration standard in the determination of elements in rainwater using Inductively Coupled Plasma Mass Spectrometer (ICP-MS).

Packaging

500ml in HD-PE bottle

Analysis Note

Al (aluminium): 50 mg/l
As (arsenic): 50 mg/l
Ba (barium): 50 mg/l
Cd (cadmium): 50 mg/l
Co (cobalt): 50 mg/l
Cr (chromium): 50 mg/l
Cu (copper): 50 mg/l
Mn (manganese): 50 mg/l
Mo (molybdenum): 50 mg/l
Ni (nickel): 50 mg/l
Pb (lead): 50 mg/l
Se (selenium): 50 mg/l
Sr (strontium): 50 mg/l
Zn (zinc): 50 mg/l
K (potassium): 500 mg/l

Other Notes

For a complete product listing of our Certipur® range of CRMs for ICP and AAS, technical information, and example certificates please visit our ICP & AAS standards website
For a complete product listing of our ICP and AAS range, including technical information and example certificates, please visit ICP & AAS standards website. 


For detailed information for our newly launched ICP-Multi element Standards check the flyer: Commitment to Quality: Elevating Certipur® Standards to Certified Reference Material   

In our effort to be more sustainable, we′re going digital with COAs-download them directly from our website.

Legal Information

Certipur is a registered trademark of Merck KGaA, Darmstadt, Germany
CERTIPUR is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service