Skip to Content
Merck
All Photos(1)

Key Documents

07-598

Sigma-Aldrich

Anti-GluR2/3 Antibody

Upstate®, from rabbit

Synonym(s):

Anti-GluA2, Anti-GluR-2, Anti-GluR-B, Anti-GluR-K2, Anti-GluR2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41
Pricing and availability is not currently available.

biological source

rabbit

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

species reactivity

rat, human, chicken, monkey, mouse

manufacturer/tradename

Upstate®

technique(s)

immunoprecipitation (IP): suitable
western blot: suitable

isotype

IgG

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

human ... GRIA2(2891)

Specificity

Recognizes GluR2/3.

Immunogen

synthetic peptide corresponding to amino acids 864-883 of rat GluR2

Application

Anti-GluR2/3 Antibody detects level of GluR2/3 & has been published & validated for use in IP & WB.
Research Category
Neuroscience
Research Sub Category
Neurotransmitters & Receptors

Neurodegenerative Diseases

Quality

routinely evaluated by immunoblot on rat brain microsomal preparation (Catalog # 12-144)

Target description

110 kDa

Physical form

Format: Purified
Protein A Purified immunoglobulin in 30% glycerol, 0.07M Tris-glycine, pH 7.4, 0.105 M NaCl, 0.035% sodium azide as a preservative.
Protein A purified

Storage and Stability

Maintain for 2 years at -20°C from date of shipment. Aliquot to avoid repeated freezing and thawing. For maximum recovery of product, centrifuge the original vial after thawing and prior to removing the cap.

Analysis Note

Control
Mouse brain tissue, rat brain microsomal preparation, human brain lysates

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A critical role for protein degradation in the nucleus accumbens core in cocaine reward memory.
Ren, ZY; Liu, MM; Xue, YX; Ding, ZB; Xue, LF; Zhai, SD; Lu, L
Neuropsychopharmacology null
Jessica A Panzer et al.
Journal of neuroimmunology, 286, 86-92 (2015-08-25)
Opsoclonus myoclonus ataxia syndrome (OMAS) is an autoimmune disorder characterized by rapid, random, conjugate eye movements (opsoclonus), myoclonus, and ataxia. Given these symptoms, autoantibodies targeting the cerebellum or brainstem could mediate the disease or be markers of autoimmunity. In a
Antigenic and mechanistic characterization of anti-AMPA receptor encephalitis.
Gleichman, AJ; Panzer, JA; Baumann, BH; Dalmau, J; Lynch, DR
Annals of Clinical and Translational Neurology null
Wei-Li Zhu et al.
Journal of neurochemistry, 117(6), 1075-1086 (2011-04-27)
Calcineurin is a serine/threonine protein phosphatase that regulates neurotransmission, neuronal structure and plasticity, and neuronal excitability in mood disorders, including depression. Increasing evidence has suggested that calcineurin is involved in the regulation of depressive-like behavior. However, little is known about
Mechanisms of seizure-induced 'transcriptional channelopathy' of hyperpolarization-activated cyclic nucleotide gated (HCN) channels.
Cristina Richichi,Amy L Brewster,Roland A Bender,Timothy A Simeone,Qinqin Zha,Hong Z Yin et al.
Neurobiology of Disease null

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service