Skip to Content
Merck
All Photos(1)

Key Documents

05-246

Sigma-Aldrich

Anti-Integrin α1 Antibody, clone 5E8D9

clone 5E8D9, Upstate®, from mouse

Synonym(s):

Anti-CD49a, Anti-VLA1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41
clone:
5E8D9, monoclonal
application:
ICC
Neutral
species reactivity:
human
technique(s):
immunocytochemistry: suitable
neutralization: suitable
citations:
15

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

5E8D9, monoclonal

species reactivity

human

manufacturer/tradename

Upstate®

technique(s)

immunocytochemistry: suitable
neutralization: suitable

isotype

IgG2a

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

human ... ITGA1(3672)

Specificity

integrinα1

Immunogen

Purified human integrinα1

Application

Anti-Integrin α1 Antibody, clone 5E8D9 is an antibody against Integrin α1 for use in IC, NEUT.
Research Category
Cell Structure
Research Sub Category
Integrins

Quality

routinely evaluated on cold ethanol fixed cells in immunocytochemistry

Target description

150kDa

Physical form

0.1M Tris-glycine, pH 7.4, 0.15M NaCl
Format: Purified
Protein A chromatography

Storage and Stability

2 years at -20°C

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Pro-adhesive and chemotactic activities of thrombospondin-1 for breast carcinoma cells are mediated by alpha3beta1 integrin and regulated by insulin-like growth factor-1 and CD98.
S Chandrasekaran, N H Guo, R G Rodrigues, J Kaiser, D D Roberts
The Journal of Biological Chemistry null
S Makihira et al.
The Journal of biological chemistry, 274(16), 11417-11423 (1999-04-10)
Cartilage matrix protein (CMP; also known as matrilin-1), one of the major noncollagenous proteins in most cartilages, binds to aggrecan and type II collagen. We examined the effect of CMP on the adhesion of chondrocytes and fibroblasts using CMP-coated dishes.
D A Calderwood et al.
The Journal of biological chemistry, 272(19), 12311-12317 (1997-05-09)
The integrin alpha1beta1 is a cell surface receptor for collagens and laminin. The alpha1 subunit contains an A-domain, and the A-domains of other integrins are known to mediate ligand binding. To determine the role of the alpha1 A-domain in ligand
Shaohua Li et al.
The American journal of pathology, 163(3), 1045-1056 (2003-08-26)
Assembly of collagen into fibrils is widely studied as a spontaneous and entropy-driven process. To determine whether vascular smooth muscle cells (SMCs) impact the formation of collagen fibrils, we microscopically tracked the conversion of soluble to insoluble collagen in human
Regulation of polymorphonuclear leukocyte cytokine receptor expression: the role of altered oxygen tensions and matrix proteins
Simms, H. and D'Amico, R.
Journal of immunology (Baltimore, Md. : 1950), 157, 3605-3616 (1996)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service