Skip to Content
Merck
All Photos(3)

Key Documents

04-958

Sigma-Aldrich

Anti-Beta Catenin Antibody, clone 7F7.2

clone 7F7.2, Upstate®, from mouse

Synonym(s):

Beta Catenin, Catenin (cadherin-associated protein), beta 1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41
clone:
7F7.2, monoclonal
application:
ICC
WB
species reactivity:
rat, mouse, human
technique(s):
immunocytochemistry: suitable
western blot: suitable
citations:
7

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

7F7.2, monoclonal

species reactivity

rat, mouse, human

manufacturer/tradename

Upstate®

technique(s)

immunocytochemistry: suitable
western blot: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... CTNNB1(1499)
mouse ... Ctnnb1(12387)
rat ... Ctnnb1(84353)

General description

Catenins are proteins found in complexes with cadherin cell adhesion molecules of animal cells. The first two catenins that were identified became known as α-catenin and β-catenin. α-catenin can bind to β-catenin and can also bind actin. β-catenin binds the cytoplasmic domain of some cadherins. When β-catenin was sequenced it was found to be a member of the armadillo family of proteins. These proteins have multiple copies of the so-called armadillo repeat domain which is specialized for protein-protein binding. An increase in β-catenin production has been noted in those people who have Basal Cell Carcinoma and leads to the increase in proliferation of related tumors. When β-catenin is not associated with cadherins and α-catenin, it can interact with other proteins such as Catenin Beta Interacting Protein 1 (ICAT) and adenomatosis polyposis coli (APC). Recent evidence suggests that β-catenin plays an important role in various aspects of liver biology including liver development (both embryonic and postnatal), liver regeneration following partial hepatectomy, HGF-induced hepatpomegaly, liver zonation, and pathogenesis of liver cancer.

Specificity

This antibody recognizes beta catenin.

Immunogen

Full length, GST-tagged recombinant beta catenin.

Application

Western Blot Analysis: 0.5-2 µg/mL of this lot detected beta catenin in HEK293 cell lysate.
Research Category
Signaling
Research Sub Category
Developmental Signaling
This Anti-Beta Catenin Antibody, clone 7F7.2 is validated for use in WB, IC for the detection of Beta Catenin.

Quality

Routinely evaluated by western blot on HEK293 cell lysate.

Target description

92 kDa

Linkage

Replaces: 04-1002

Physical form

Format: Purified
Protein G Purified
Purified mouse monoclonal IgG1κ in buffer containing 0.1 M Tris-Glycine, pH 7.4, 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
HEK293 cell lysate, PC12 cell lysate, NIH/3T3 cell lysate

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Penelope M Tsimbouri et al.
Nature biomedical engineering, 1(9), 758-770 (2017-09-01)
Bone grafts are one of the most commonly transplanted tissues. However, autologous grafts are in short supply, and can be associated with pain and donor-site morbidity. The creation of tissue-engineered bone grafts could help to fulfil clinical demand and provide
Na Li et al.
International journal of molecular sciences, 17(4) (2016-04-23)
R-spondin proteins are novel Wnt/β-catenin agonists, which signal through their receptors leucine-rich repeat-containing G-protein coupled receptor (LGR) 4/5/6 and substantially enhance Wnt/β-catenin activity. R-spondins are reported to function in embryonic development. They also play important roles in stem cell functions
Lei Wang et al.
Oncology reports, 42(5), 2169-2178 (2019-08-23)
Esophageal adenocarcinoma (EAC) is an aggressive and challenging disease to treat, with an overall five‑year survival rate of <20%. Early malignant cell dissemination contributes to this poor prognosis. Epithelial‑mesenchymal transition (EMT) induces the invasion and metastasis of carcinoma cells. Empty
Wei Feng et al.
Molecular medicine reports, 12(3), 4727-4733 (2015-06-23)
Previous studies have demonstrated that the Wnt/β‑catenin signaling pathway is critical to the induction and maintenance of chronic neuropathic pain caused by peripheral inflammation and nerve damage. Emerging evidence from recent studies suggests that epigenetic mechanisms may also be critical
Miriam Mecha et al.
Journal of neuroinflammation, 17(1), 88-88 (2020-03-21)
The participation of microglia in CNS development and homeostasis indicate that these cells are pivotal for the regeneration that occurs after demyelination. The clearance of myelin debris and the inflammatory-dependent activation of local oligodendrocyte progenitor cells in a demyelinated lesion

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service