Skip to Content
Merck
All Photos(1)

Key Documents

R-026

Supelco

Remifentanil acid solution

100 μg/mL in acetonitrile, certified reference material, Cerilliant®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H26N2O5
CAS Number:
Molecular Weight:
362.42
UNSPSC Code:
41116107
NACRES:
NA.24
Pricing and availability is not currently available.

grade

certified reference material

Quality Level

form

liquid

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

100 μg/mL in acetonitrile

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

forensics and toxicology

format

single component solution

storage temp.

−20°C

SMILES string

N1(CCC(CC1)(N(c2ccccc2)C(=O)CC)C(=O)OC)CCC(=O)O

InChI

1S/C19H26N2O5/c1-3-16(22)21(15-7-5-4-6-8-15)19(18(25)26-2)10-13-20(14-11-19)12-9-17(23)24/h4-8H,3,9-14H2,1-2H3,(H,23,24)

InChI key

GFJKFSFFMHOISI-UHFFFAOYSA-N

General description

Remifentanil acid is the major urinary metabolite of remifentanil, a synthetic opioid used to induce or supplement general anesthesia. This new certified reference solution is suitable for a variety of mass spectrometry-based testing applications including clinical toxicology, forensic analysis, workplace drug testing and pharmaceutical research.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

related product

Product No.
Description
Pricing

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

35.6 °F - closed cup

Flash Point(C)

2.0 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service