Skip to Content
Merck
All Photos(1)

Key Documents

LM2000

Avanti

Sphingosine (d17:1)

Avanti Research - A Croda Brand LM2000, ethanol solution

Synonym(s):

heptadeca-sphing-4-enine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H35NO2
CAS Number:
Molecular Weight:
285.47
UNSPSC Code:
12352211
NACRES:
NA.25

form

ethanol solution

packaging

pkg of 1 × 1 mL (LM2000-1EA)

manufacturer/tradename

Avanti Research - A Croda Brand LM2000

concentration

~10 μg/mL (Refer to C of A for lot specific concentration.)

shipped in

dry ice

storage temp.

−20°C

SMILES string

OC[C@@](N)([H])[C@]([H])(O)/C=C/CCCCCCCCCCCC

InChI

1S/C17H35NO2/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-17(20)16(18)15-19/h13-14,16-17,19-20H,2-12,15,18H2,1H3/b14-13+/t16-,17+/m0/s1

InChI key

RBEJCQPPFCKTRZ-LHMZYYNSSA-N

Application

Sphingosine (d17:1) or heptadeca-sphing-4-enine might be used as an internal standard for the identification of long chain bases (LCBs) in Colletotrichum gloeosporioides mycelia. It is also used as an internal standard for sample preparation and LC-MS/MS analysis for sphingolipids.

Packaging

2 mL Amber Glass Sealed Ampule (LM2000-1EA)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

55.4 °F - closed cup

Flash Point(C)

13.0 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yimei Huang et al.
Frontiers in microbiology, 10, 1016-1016 (2019-06-06)
Fungal glucosylceramide plays important role in cell division, hyphal formation and growth, spore germination and the modulation of virulence and has recently been considered as target for small molecule inhibitors. In this study, we characterized CgGCS, a protein encoding a
Claudia Rossi et al.
International journal of molecular sciences, 19(11) (2018-11-18)
Multiple sclerosis (MuS) is an autoimmune disease of the central nervous system characterized by neuroinflammation, neurodegeneration, and degradation of the myelin sheath. Epidemiological studies have shown that the female gender is more susceptible than the male gender to MuS development
Veera Venkata Ratnam Bandaru et al.
BMC neuroscience, 15, 137-137 (2014-12-30)
Cholesterol metabolism is important for the maintenance of myelin and neuronal membranes in the central nervous system. Blood concentrations of the brain specific cholesterol metabolite 24S-hydroxysterol to the peripheral metabolite 27-hydroxycholesterol may be useful surrogate markers for neurodegenerative diseases including
Christopher M Adams et al.
The Journal of biological chemistry, 279(50), 52772-52780 (2004-09-29)
The current paper demonstrates that cholesterol and its hydroxylated derivative, 25-hydroxycholesterol (25-HC), inhibit cholesterol synthesis by two different mechanisms, both involving the proteins that control sterol regulatory element-binding proteins (SREBPs), membrane-bound transcription factors that activate genes encoding enzymes of lipid
Siker Kimbung et al.
Endocrine-related cancer, 24(7), 339-349 (2017-04-27)
The impact of systemic 27-hydroxycholesterol (27HC) and intratumoral CYP27A1 expression on pathobiology and clinical response to statins in breast cancer needs clarification. 27HC is an oxysterol produced from cholesterol by the monooxygenase CYP27A1, which regulates intracellular cholesterol homeostasis. 27HC also

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service