Skip to Content
Merck
All Photos(1)

Key Documents

890808P

Avanti

22:0 Trehalose

Avanti Research - A Croda Brand

Synonym(s):

D-(+)-trehalose 6,6′-dibehenate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C56H106O13
CAS Number:
Molecular Weight:
987.43
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25

form

powder

packaging

pkg of 1 × 25 mg (890808P-25mg)
pkg of 1 × 5 mg (890808P-5mg)

manufacturer/tradename

Avanti Research - A Croda Brand

application(s)

vaccine development

shipped in

dry ice

storage temp.

−20°C

SMILES string

O[C@H]([C@@H](COC(CCCCCCCCCCCCCCCCCCCCC)=O)O[C@H](O[C@@H](O[C@@H]([C@H]([C@@H]1O)O)COC(CCCCCCCCCCCCCCCCCCCCC)=O)[C@@H]1O)[C@@H]2O)[C@@H]2O

InChI key

ZLJJDBSDZSZVTF-LXOQPCSCSA-N

General description

22:0 Trehalose is a bioactive glycolipid and an analog of mycobacterial cord factor.

Application

22:0 Trehalose has been used as an adjuvant for Chlamydia vaccine. It has also been used as a component in adjuvant DMT (dimethyldioctadecylammonium/monophosphoryl lipid A/trehalose 6,6′-dibehenate (DDA/MPL/TDB)) to prepare DMT liposome adjuvanted CTT3H (polyprotein) vaccine.

Biochem/physiol Actions

Incorporation of trehalose 6,6′-dibehenate (TDB) into cationic liposomes composed of the quaternary ammonium compound dimethyldioctadecylammonium (DDA) produce an adjuvant system which induces a powerful cell-mediated immune response and a strong antibody response, desirable for a high number of disease targets. It acts as an immunomodulator. 22:0 Trehalose is considered less toxic and an efficient vaccine adjuvant for tuberculosis.

Packaging

5 mL Amber Glass Screw Cap Vial (890808P-25mg)
5 mL Amber Glass Screw Cap Vial (890808P-5mg)

Other Notes

For R&D use only. Not for drug, household, or other uses.

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hong Yu et al.
Infection and immunity, 78(5), 2272-2282 (2010-03-17)
Major impediments to developing a Chlamydia vaccine lie in identifying immunologically relevant T-cell antigens and delivery in a manner to stimulate protective immunity. Using an immunoproteomic approach, we previously identified three immunodominant Chlamydia T-cell antigens (PmpG-1, PmpE/F-2, and RplF). Because
Xindong Teng et al.
Human vaccines & immunotherapeutics, 11(6), 1456-1464 (2015-04-24)
Different strategies have been proposed for the development of protein subunit vaccine candidates for tuberculosis (TB), which shows better safety than other types of candidates and the currently used Bacillus Calmette-Guérin (BCG) vaccine. In order to develop more effective protein

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service