Skip to Content
Merck
All Photos(1)

Key Documents

880001M

Avanti

Chol-PEG600

Avanti Research - A Croda Brand 880001M, methylene chloride solution

Synonym(s):

Cholesterol-(polyethylene glycol-600)

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25

form

methylene chloride solution

packaging

pkg of 1 × 2.5 mL (880001M-25mg)

manufacturer/tradename

Avanti Research - A Croda Brand 880001M

concentration

10 mg/mL (880001M-25mg)

shipped in

dry ice

storage temp.

−20°C

SMILES string

O(C1CCC2(C3C(C4C(C(CC4)C(CCCC(C)C)C)(CC3)C)CC=C2C1)C)CCO

InChI

1S/C29H50O2/c1-20(2)7-6-8-21(3)25-11-12-26-24-10-9-22-19-23(31-18-17-30)13-15-28(22,4)27(24)14-16-29(25,26)5/h9,20-21,23-27,30H,6-8,10-19H2,1-5H3

InChI key

NPYXBHZQVNRAOF-UHFFFAOYSA-N

General description

Chol-PEG600, also known as cholesterol-(polyethylene glycol-600), is a derivative of cholesterol and is a nonionic amphipathic lipid. Structurally, it comprises lipophilic cholesterol and covalently bound hydrophilic PEG.

Biochem/physiol Actions

PEG-cholesterol is mainly used in preparation of lipoplexes for DNA transfection studies, and liposomes for encapsulated drug delivery.

Packaging

5 mL Clear Glass Sealed Ampule (880001M-25mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Carc. 2 - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Central nervous system

WGK

WGK 2


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Poly (ethylene glycol)-cholesterol inhibits L-type Ca2+ channel currents and augments voltage-dependent inactivation in A7r5 cells
Ochi, R, et al.
Testing, 9(9) (2014)
Rikuo Ochi et al.
PloS one, 9(9), e107049-e107049 (2014-09-10)
Cholesterol distributes at a high density in the membrane lipid raft and modulates ion channel currents. Poly(ethylene glycol) cholesteryl ether (PEG-cholesterol) is a nonionic amphipathic lipid consisting of lipophilic cholesterol and covalently bound hydrophilic PEG. PEG-cholesterol is used to formulate
CHOBIMALT: a cholesterol-based detergent
Howell S, et al.
Biochemistry, 49(44), 9572-9583 (2010)
Poly (ethylene glycol)-cholesterol inhibits L-type Ca2+ channel currents and augments voltage-dependent inactivation in A7r5 cells
Ochi, R, et al.
PLoS ONE, 9(9) (2014)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service