Skip to Content
Merck
All Photos(1)

Key Documents

860629P

Avanti

C24:1 Dihydroceramide (d18:0/24:1(15Z))

Avanti Research - A Croda Brand 860629P, powder

Synonym(s):

N-nervonoyl-D-erythro-sphinganine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C42H83NO3
CAS Number:
Molecular Weight:
650.11
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25

form

powder

packaging

pkg of 1 × 5 mg (860629P-5mg)

manufacturer/tradename

Avanti Research - A Croda Brand 860629P

lipid type

sphingolipids

shipped in

dry ice

storage temp.

−20°C

SMILES string

OC[C@]([H])(NC(CCCCCCCCCCCCC/C=C\CCCCCCCC)=O)[C@]([H])(O)CCCCCCCCCCCCCCC

InChI

1S/C42H83NO3/c1-3-5-7-9-11-13-15-17-18-19-20-21-22-23-24-26-28-30-32-34-36-38-42(46)43-40(39-44)41(45)37-35-33-31-29-27-25-16-14-12-10-8-6-4-2/h17-18,40-41,44-45H,3-16,19-39H2,1-2H3,(H,43,46)/b18-17-/t40-,41+/m0/s1

InChI key

YUULKFVZRXQHPM-ATHUGRIKSA-N

Application

C24:1 Dihydroceramide has been used as a standard in quadrupole time-of-flight (Q-TOF) mass spectrometry for the measurement of sphingolipid metabolites in adipose tissue. It has also been used a standard in diacylglycerol kinase (DGK) assay to quantify dihydroceramides in CLN9-deficient cells.

Biochem/physiol Actions

C24:1 Dihydroceramide is a synthetic dihydroceramide. Dihydroceramides are reduced to ceramides by the enzyme dihydroceramide desaturase. Elevated levels of serum 24:1 DHC has been observed in patients with history of hepatocellular carcinoma (HCC) pre-orthopic liver transplantation (OLT), compared to patients with no prior HCC. Therefore, 24:1 DHC can be used as a HCC biomarker in post-OLT patients.

Packaging

5 mL Amber Glass Screw Cap Vial (860629P-5mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

C22: 0-and C24: 0-dihydroceramides confer mixed cytotoxicity in T-cell acute lymphoblastic leukemia cell lines
Holliday Jr, et al.
Testing, 8(9) (2013)
C22: 0-and C24: 0-dihydroceramides confer mixed cytotoxicity in T-cell acute lymphoblastic leukemia cell lines
Holliday Jr, et al.
PLoS ONE, 8(9) (2013)
The CLN9 protein, a regulator of dihydroceramide synthase
Schulz A, et al.
The Journal of Biological Chemistry, 281(5), 2784-2794 (2006)
Michael W Holliday et al.
PloS one, 8(9), e74768-e74768 (2013-09-17)
We previously reported that fenretinide (4-HPR) was cytotoxic to acute lymphoblastic leukemia (ALL) cell lines in vitro in association with increased levels of de novo synthesized dihydroceramides, the immediate precursors of ceramides. However, the cytotoxic potentials of native dihydroceramides have
Angela Schulz et al.
The Journal of biological chemistry, 281(5), 2784-2794 (2005-11-24)
A new variant of a group of pediatric neurodegenerative diseases known as neuronal ceroid lipofuscinosis (NCL) or Batten disease has been identified. It is termed CLN9-deficient. CLN9-deficient fibroblasts have a distinctive phenotype of rapid growth and increased apoptosis and diminished

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service