Skip to Content
Merck
All Photos(1)

Key Documents

860056P

Avanti

Ganglioside GD1b

powder, Avanti Research - A Croda Brand (60056P)

Synonym(s):

Ganglioside GD1b (porcine brain, triammonium salt)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C84H154N6O39
CAS Number:
Molecular Weight:
1872.14
UNSPSC Code:
12352211
NACRES:
NA.25

Assay

99% (TLC)

form

powder

packaging

pkg of 1 × 1 mg (860056P-1MG)

manufacturer/tradename

Avanti Research - A Croda Brand (60056P)

lipid type

sphingolipids
lipid extracts

shipped in

dry ice

storage temp.

−20°C

SMILES string

[H][C@](/C=C/CCCCCCCCCCCCC)(O)[C@@]([H])(NC(CCCCCCCCCCCCCCCCC)=O)CO[C@H]1[C@H](O)[C@@H](O)[C@H](O[C@@H]2O[C@H](CO)[C@@H]([C@H](O[C@@]3(C([O-])=O)O[C@@H]([C@H](O)[C@H](O[C@@]4(C([O-])=O)O[C@@H]([C@H](O)[C@H](O)CO)[C@H](NC(C)=O)[C@@H](O)C4)CO)[C@@](NC(C)=O)

General description

Ganglioside GD1b is a complex brain ganglioside. It is also present in the serum and are transported by the serum lipoproteins.

Application

Ganglioside GD1b has been used in the purification of GD1b (d20:1−20:0) and GD1b (d20:1−18:0) by high performance liquid chromatography (HPLC) method.

Biochem/physiol Actions

Ganglioside GD1b functions as a receptor for BK virus.

Packaging

5 mL Amber Glass Screw Cap Vial

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Imbalance in fatty-acid-chain length of gangliosides triggers Alzheimer amyloid deposition in the precuneus
Oikawa N, et al.
PLoS ONE, 10(3) (2015)
Ganglioside biochemistry
Kolter T.
ISRN Biochemistry, 2012(10), 537-544 (2012)
Imbalance in fatty-acid-chain length of gangliosides triggers Alzheimer amyloid deposition in the precuneus
Oikawa N, et al.
Testing, 10(3) (2015)
Identification of gangliosides GD1b and GT1b as receptors for BK virus
Low J A, et al.
Journal of virology, 80(3), 1361-1366 (2006)
Structures, biosynthesis, and functions of gangliosides-an overview
Robert K, et al.
Journal of Oleo Science, 60(10), 537-544 (2011)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service