Skip to Content
Merck
All Photos(2)

Key Documents

855775C

Avanti

18:0 Lyso PC

1-stearoyl-2-hydroxy-sn-glycero-3-phosphocholine, chloroform

Synonym(s):

1-octadecanoyl-sn-glycero-3-phosphocholine; PC(18:0/0:0)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C26H54NO7P
CAS Number:
Molecular Weight:
523.68
MDL number:
UNSPSC Code:
51191904
NACRES:
NA.25
Pricing and availability is not currently available.

Assay

>99% (LPC; may contain up to 10% of the 2-LPC isomer, TLC)

form

liquid

packaging

pkg of 1 × 2.5 mL (855775C-25mg)
pkg of 1 × 20 mL (855775C-500mg)

manufacturer/tradename

Avanti Research - A Croda Brand 855775C

concentration

10 mg/mL (855775C-25mg)
25 mg/mL (855775C-500mg)

shipped in

dry ice

storage temp.

−20°C

SMILES string

[P](=O)([O-])(OCC[N+](C)(C)C)OCC(O)COC(=O)CCCCCCCCCCCCCCCCC

InChI

1S/C26H54NO7P/c1-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19-20-26(29)32-23-25(28)24-34-35(30,31)33-22-21-27(2,3)4/h25,28H,5-24H2,1-4H3

InChI key

IHNKQIMGVNPMTC-UHFFFAOYSA-N

Application

18:0 Lyso PC has been used in gametocytogenesis assay.[1]

Packaging

30 mL Amber Narrow Mouth Glass Bottle with Screw Cap (855775C-500mg)
5 mL Clear Glass Sealed Ampule (855775C-25mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 3 - Carc. 2 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 - STOT SE 3

Target Organs

Central nervous system, Liver,Kidney

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lysophosphatidylcholine regulates sexual stage differentiation in the human malaria parasite Plasmodium falciparum
Brancucci NMB, et al.
Cell, 171(7), 1532-1544 (2017)
Anthony J Valente et al.
Free radical biology & medicine, 70, 117-128 (2014-02-25)
Oxidized low-density lipoprotein (oxLDL) induces endothelial cell death through the activation of NF-κB and AP-1 pathways. TRAF3IP2 is a redox-sensitive cytoplasmic adapter protein and an upstream regulator of IKK/NF-κB and JNK/AP-1. Here we show that oxLDL-induced death in human primary
Dumindika A Siriwardane et al.
International journal of pharmaceutics, 578, 119077-119077 (2020-01-29)
Identification and quantification of excipient related degradation products in the liposomal formulation is important, as they may impact the safety and efficacy of the drug. Phospholipids are one of the major excipients in liposome drugs composing the lipid bilayer, and
Danielle M Charron et al.
Langmuir : the ACS journal of surfaces and colloids, 36(19), 5385-5393 (2020-04-28)
Porphyrin aggregates have attractive photophysical properties for phototherapy and optical imaging, including quenched photosensitization, efficient photothermal conversion, and unique absorption spectra. Although hydrophobic porphyrin photosensitizers have long been encapsulated into liposomes for drug delivery, little is known about the membrane
Otto K Kari et al.
Nanoscale, 12(3), 1728-1741 (2020-01-03)
Methodological constraints have limited our ability to study protein corona formation, slowing nanomedicine development and their successful translation into the clinic. We determined hard and soft corona structural properties along with the corresponding proteomic compositions on liposomes in a label-free

Articles

The critical micelle concentration (CMC) can be approximately defined as the lipid monomer concentration at which appreciable amounts (>5% of total) of micellar aggregates first begin to appear in the equilibrium: nM1<=>Mn

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service