Skip to Content
Merck
All Photos(2)

Key Documents

850385C

Avanti

18:2 (Cis) PC (DLPC)

1,2-dilinoleoyl-sn-glycero-3-phosphocholine, chloroform

Synonym(s):

1,2-di-(9Z,12Z-octadecadienoyl)-sn-glycero-3-phosphocholine; PC(18:2(9Z,12Z)/18:2(9Z,12Z))

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C44H80NO8P
CAS Number:
Molecular Weight:
782.08
UNSPSC Code:
51191904
NACRES:
NA.25

Assay

>99% (TLC)

form

liquid

packaging

pkg of 1 × 2.5 mL (850385C-25mg)
pkg of 2 × 4 mL (850385C-200mg)

manufacturer/tradename

Avanti Research - A Croda Brand 850385C

concentration

10 mg/mL (850385C-25mg)
25 mg/mL (850385C-200mg)

lipid type

cardiolipins
phospholipids

shipped in

dry ice

storage temp.

−20°C

InChI

1S/C44H80NO8P/c1-6-8-10-12-14-16-18-20-22-24-26-28-30-32-34-36-43(46)50-40-42(41-52-54(48,49)51-39-38-45(3,4)5)53-44(47)37-35-33-31-29-27-25-23-21-19-17-15-13-11-9-7-2/h14-17,20-23,42H,6-13,18-19,24-41H2,1-5H3/b16-14-,17-15-,22-20-,23-21-/t42-/m1/s1

InChI key

FVXDQWZBHIXIEJ-LNDKUQBDSA-N

General description

18:2 (Cis) PC (DLPC) is a synthetic phosphatidylcholine.
The list of Phosphatidylcholine products offered by Avanti is designed to provide compounds having a variety of physical properties. Products available include short chain (C3-C8 are water soluble and hygroscopic), saturated, multi-unsaturated and mixed acid PC′s. All of the products are purified by HPLC, and special precautions are taken to protect the products for oxidization and hydrolysis.

Application

18:2 (Cis) PC (DLPC) has been used to evaluate the performance of gradient laser ablation electrospray ionization (g-LAESI) for analyzing compounds with different polarities. It has also been used as a lipid standard in the optimization of the liquid chromatography-mass spectrometry (LC-MS) method conditions.

Packaging

5 mL Clear Glass Sealed Ampule (850385C-200mg)
5 mL Clear Glass Sealed Ampule (850385C-25mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

also commonly purchased with this product

Product No.
Description
Pricing

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 3 - Carc. 2 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 - STOT SE 3

Target Organs

Central nervous system, Liver,Kidney

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Comprehensive untargeted lipidomic analysis using core-shell C30 particle column and high field orbitrap mass spectrometer
Narvaez-Rivas M and Zhang Q
Journal of Chromatography A, 1440(16), 123-134 (2016)
Yueheng Zhang et al.
Langmuir : the ACS journal of surfaces and colloids, 33(11), 2780-2789 (2017-03-02)
We report the ability of hydrophobically modified polypeptoids (HMPs), which are amphiphilic pseudopeptidic macromolecules, to connect across lipid bilayers and thus form layered structures on liposomes. The HMPs are obtained by attaching hydrophobic decyl groups at random points along the
Solvent gradient electrospray for laser ablation electrospray ionization mass spectrometry
Li H and Vertes A
Analyst, 142(16), 2921-2927 (2017)
Xiao Wu et al.
International journal of pharmaceutics, 579, 119168-119168 (2020-02-23)
Beta-lapachone (β-Lap) is an anticancer drug activated by the NAD(P)H:quinone oxidoreductase (NQO1), an enzyme over-expressed in a large variety of tumors. B-Lap is poorly soluble in water and in most biocompatible solvents. Micellar systems, liposomes and cyclodextrins (CDs) have been
Mariana Vignoni et al.
Langmuir : the ACS journal of surfaces and colloids, 34(50), 15578-15586 (2018-11-21)
Pterins are natural products that can photosensitize the oxidation of DNA, proteins, and phospholipids. Recently, a new series of decyl-chain (i.e., lipophilic) pterins were synthesized and their photophysical properties were investigated. These decyl-pterins led to efficient intercalation in large unilamellar

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service