Skip to Content
Merck
All Photos(3)

Key Documents

850081P

Avanti

Monolyso Heart CA

Avanti Research - A Croda Brand 850081P, powder

Synonym(s):

monolysocardiolipin (Heart, Bovine) (sodium salt)

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
UNSPSC Code:
12352211
NACRES:
NA.25
Pricing and availability is not currently available.

Assay

>99% (TLC)

form

powder

packaging

pkg of 1 × 10 mg (850081P-10mg)

manufacturer/tradename

Avanti Research - A Croda Brand 850081P

lipid type

phosphoglycerides

shipped in

dry ice

storage temp.

−20°C

SMILES string

O=P([O-])(OC[C@]([H])(O)COC(CCCCCCC/C=C\C/C=C\CCCCC)=O)OCC([H])(O)COP([O-])(OC[C@]([H])(OC(CCCCCCC/C=C\C/C=C\CCCCC)=O)COC(CCCCCCC/C=C\C/C=C\CCCCC)=O)=O.[Na+].[Na+]

InChI key

BYCQHLJVUPZGJA-LWABKDCLSA-L

General description

Monolysocardiolipin (MLCL) is a class of phospholipid related to cardiolipin.[1] MLCL is a triacylated phospholipid, uncommon in bacterial or eukaryotic membranes.[2] MLCL is prepared from cardiolipin by the hydrolysis action of phospholipase A2 (PLA2).[3]

Application

Monolyso Heart CA is suitable for use:
  • as substrate for the synthesis of cardiolipin by the enzyme acyl-CoA:lysocardiolipin acyltransferase (ACLAT1)[4]
  • as substrate for the assay of mitochondrial monolysocardiolipin acyltransferase (MLCL AT) activity[5]
  • in the preparation of affinity resin MLCL-adriamycin-agarose for the purification of MLCL AT[6]

Biochem/physiol Actions

Monolysocardiolipin (MLCL) is a substrate for the biosynthesis of cardiolipin in heart by the activity of MLCL acyltransferase.[5] MLCL is a marker for Barth syndrome.[1] MLCL is implicated in apoptosis.[3]

Packaging

5 mL Clear Glass Sealed Ampule (850081P-10mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Identification of unique cardiolipin and monolysocardiolipin species in Acinetobacter baumannii
Lopalco P, et al.
Scientific Reports, 7(1), 2972-2972 (2017)
Monolysocardiolipin: improved preparation with high yield
Kim J and Hoppel CL
Journal of Lipid Research, 52(2), 389-392 (2011)
Thyroxine regulation of monolysocardiolipin acyltransferase activity in rat heart
Mutter T, et al.
The Biochemical Journal, 346(2), 403-406 (2000)
Identification of the human mitochondrial linoleoyl-coenzyme A monolysocardiolipin acyltransferase (MLCL AT-1)
Taylor WA and Hatch GM
The Journal of Biological Chemistry, 284(44), 30360-30371 (2009)
Monolysocardiolipin in cultured fibroblasts is a sensitive and specific marker for Barth Syndrome
Van Werkhoven MA, et al.
Journal of Lipid Research, 47(10), 2346-2351 (2006)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service