Skip to Content
Merck
All Photos(2)

Key Documents

840419P

Avanti

16:0 Phosphatidylmethanol

1,2-dipalmitoyl-sn-glycero-3-phosphomethanol (sodium salt), powder

Synonym(s):

1,2-dihexadecanoyl-sn-glycero-3-phosphomethanol (sodium salt)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C36H70O8PNa
CAS Number:
Molecular Weight:
684.90
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25

Assay

>99% (TLC)

form

powder

packaging

pkg of 1 × 25 mg (840419P-25mg)

manufacturer/tradename

Avanti Research - A Croda Brand 840419P

shipped in

dry ice

storage temp.

−20°C

SMILES string

[H][C@@](COP([O-])(OC)=O)(OC(CCCCCCCCCCCCCCC)=O)COC(CCCCCCCCCCCCCCC)=O.[Na+]

InChI

1S/C36H71O8P.Na/c1-4-6-8-10-12-14-16-18-20-22-24-26-28-30-35(37)42-32-34(33-43-45(39,40)41-3)44-36(38)31-29-27-25-23-21-19-17-15-13-11-9-7-5-2;/h34H,4-33H2,1-3H3,(H,39,40);/q;+1/p-1/t34-;/m1./s1

InChI key

JDEIVNUSWULHEB-MDYNBEAQSA-M

Application

16:0 Phosphatidylmethanol (1,2-dipalmitoyl-sn-glycero-3-phosphomethanol (sodium salt)) may be used as a standard in ultra-high performance liquid chromatography tandem mass spectrometry (UHPLC-MS/MS) analysis for the quantification of phosphatidylethanols (PEths) in whole blood, venous/capillary-dried blood spots (V-DBS and C-DBS).

Biochem/physiol Actions

Phosphatidylmethanol is an anionic lipid which binds to the Phospholipase D. It is categorized as an alkyl phosphate lipid under headgroup modified lipids.

Packaging

5 mL Amber Glass Screw Cap Vial (840419P-25mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

K Stieglitz et al.
The Journal of biological chemistry, 274(50), 35367-35374 (1999-12-10)
The Streptomyces chromofuscus phospholipase D (PLD) cleavage of phosphatidylcholine in bilayers can be enhanced by the addition of the product phosphatidic acid (PA). Other anionic lipids such as phosphatidylinositol, oleic acid, or phosphatidylmethanol do not activate this PLD. This allosteric
Natalie Kummer et al.
Analytical and bioanalytical chemistry, 408(3), 825-838 (2015-11-26)
Phosphatidylethanol species (PEths) are promising biomarkers of alcohol consumption. Here, we report on the set-up, validation, and application of a novel UHPLC-ESI-MS/MS method for the quantification of PEth 16:0/18:1, PEth 18:1/18:1, and PEth 16:0/16:0 in whole blood (30 μL) and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service