Skip to Content
Merck
All Photos(1)

Key Documents

810603P

Avanti

16:0-10 Doxyl PC

Avanti Research - A Croda Brand 810603P, powder

Synonym(s):

1-palmitoyl-2-stearoyl-(10-doxyl)-sn-glycero-3-phosphocholine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C46H90N2O10P
CAS Number:
Molecular Weight:
862.19
MDL number:
UNSPSC Code:
41141825
NACRES:
NA.25

Assay

>99% (TLC)

form

powder

packaging

pkg of 1 × 1 mg (810603P-1mg)

manufacturer/tradename

Avanti Research - A Croda Brand 810603P

lipid type

ESR probes
phospholipids

shipped in

dry ice

storage temp.

−20°C

General description

Avanti′s nitroxide spin product listing is a group of compounds designed to act as membrane probes. A variety of positions down the hydrophobic chain are labeled with the nitroxide functional groups to allow probing the membrane at various depths. These compounds have been synthesized from 1-palmitoyl-2-hydroxy-sn-glycerol-3-phosphocholine with the product being purified by column chromatography. Various n-doxyl phosphocholines have been recently used as biophysical tools to elucidate membrane trafficking with phosphatidylinositol transfer proteins and as fluorescent quenchers in lipid bilayer structural studies.
Phosphocholine is considered as a precursor molecule. It is formed during the breakdown of phosphatidylcholine metabolism. 10-Doxyl is a spin label probe.

Application

16:0-10 Doxyl PC may be used:
  • in vesicles for quenching PT-(1–46)F4W fluorescence
  • as vesicle component for tryptophan based fluorescence emission studies
  • to probe the oxygen and Ni[II] ethylenediaminediacetic acid (NiEdda) accessibilities inside liposomes

Biochem/physiol Actions

Phosphatidylcholine (PC) functions as a surfactant within the mucus to form a hydrophobic surface to inhibit bacterial penetrance.

Packaging

5 mL Clear Glass Sealed Ampule (810603P-1mg)

Preparation Note

Product use: To prevent aggregation, prepare water-based solutions of 2 mM stock solutions of n-DOXYL PCs and store in plastic. Dilute stock solutions to 0.03- 0.1 mM solutions for EPR studies. For liposome preparations in fluorescent quenching measurements, dissolve the doxyl lipid in 150 μl absolute ethanol for a concentration of 40.3 mM , Additional supplemental information.

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Integrative Medicine - E-Book (2017)
Biology-cancer metabolic phenotype
NMR Metabolomics in Cancer Research, 15-138 (2013)
Qufei Li et al.
Nature structural & molecular biology, 21(2), 160-166 (2014-01-15)
Voltage-gated ion channels respond to transmembrane electric fields through reorientations of the positively charged S4 helix within the voltage-sensing domain (VSD). Despite a wealth of structural and functional data, the details of this conformational change remain controversial. Recent electrophysiological evidence
Alexander Kyrychenko et al.
The journal of physical chemistry. B, 117(19), 5875-5885 (2013-04-26)
Spin-labeled lipids are commonly used as fluorescence quenchers in studies of membrane penetration of dye-labeled proteins and peptides using depth-dependent quenching. Accurate calculations of depth of the fluorophore rely on the use of several spin labels placed in the membrane
Q Wang et al.
Biochimica et biophysica acta, 1324(1), 69-75 (1997-02-21)
The N-terminal signal sequence of glucitol permease of Escherichia coli (Gut22: MIETITPGAVWFIGLFQKGGEC) and its analog (Gut22Ana: MIETITHGAEWFIGLFQKGGEC) were synthesized. The analog had a Pro residue substituted for the His at the 7th position of Gut22 and a Val residue substituted

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service