Skip to Content
Merck
All Photos(2)

Key Documents

810123P

Avanti

14:0-12:0 NBD PC

Avanti Research - A Croda Brand 810123P, powder

Synonym(s):

1-Myristoyl-2-[12-[(7-nitro-2-1,3-benzoxadiazol-4-yl)amino]dodecanoyl]-sn-Glycero-3-Phosphocholine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C40H70N5O11P
CAS Number:
Molecular Weight:
827.98
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25

Assay

>99% (TLC)

form

powder

packaging

pkg of 1 × 1 mg (810123P-1mg)
pkg of 5 × 2 mg (810123P-10mg)

manufacturer/tradename

Avanti Research - A Croda Brand 810123P

lipid type

fluorescent lipids
phospholipids

shipped in

dry ice

storage temp.

−20°C

General description

Phosphatidylcholine (PC) is the most abundant phospholipid in eukaryotes. It is the prime metabolite of glycerolipid metabolism and a vital component of the mucosal layer of the colon. NBD PC has 7-nitrobenz-2-oxa-1,3-diazol-4-yl (NBD) moiety attached to phosphatidylcholine.

Application

14:0-12:0 NBD PC has been used to prepare fluorescently labeled 1,2-dimyristoyl-sn-glycero-3-phosphocholine (DMPC) unilamellar vesicles (LUVs) for the measurement lipid mass distribution. It may also be used to determine in vitro Phospholipase D (PLD) activity.

Biochem/physiol Actions

7-nitrobenz-2-oxa-1,3-diazol-4-yl phosphatidylcholine (NBD PC) functions as fluorescent probe involved in the study of the biological membranes in cellular processes. It is a strong bilayer-forming lipid.

Packaging

5 mL Amber Glass Screw Cap Vial (810123P-10mg)
5 mL Amber Glass Screw Cap Vial (810123P-1mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Eric Oropeza-Guzman et al.
Langmuir : the ACS journal of surfaces and colloids, 35(50), 16528-16535 (2019-11-21)
We took advantage of the microflow hydrodynamics in the evaporation of sessile droplets to increase the height uniformity of thin lipid films for the subsequent electroformation of defect-free giant unilamellar vesicles (GUV). By serially casting progressively larger liposome suspension droplets
Takeshi Yamaguchi et al.
Plant physiology, 150(1), 308-319 (2009-03-17)
Phospholipase D (PLD) plays an important role in plants, including responses to abiotic as well as biotic stresses. A survey of the rice (Oryza sativa) genome database indicated the presence of 17 PLD genes in the genome, among which OsPLDalpha1
Luís M S Loura et al.
Biochimica et biophysica acta, 1778(2), 491-501 (2007-11-21)
We present a combined theoretical (molecular dynamics, MD) and experimental (differential scanning calorimetry, DSC) study of the effect of 7-nitrobenz-2-oxa-1,3-diazol-4-yl (NBD) acyl chain-labeled fluorescent phospholipid analogs (C6-NBD-PC and C12-NBD-PC) on 1,2-dipalmitoyl-sn-glycero-3-phosphocholine (DPPC) bilayers. DSC measurements reveal that <1 mol% of
César Botella et al.
Progress in lipid research, 65, 12-23 (2016-11-23)
In plant cells, phosphatidylcholine (PC) is a major glycerolipid of most membranes but practically lacking from the plastid internal membranes. In chloroplasts, PC is absent from the thylakoids and the inner envelope membrane. It is however the main component of
Jelske N van der Veen et al.
Biochimica et biophysica acta. Biomembranes, 1859(9 Pt B), 1558-1572 (2017-04-16)
Phosphatidylcholine (PC) and phosphatidylethanolamine (PE) are the most abundant phospholipids in all mammalian cell membranes. In the 1950s, Eugene Kennedy and co-workers performed groundbreaking research that established the general outline of many of the pathways of phospholipid biosynthesis. In recent

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service