Skip to Content
Merck
All Photos(1)

Key Documents

700156P

Avanti

20α-hydroxycholesterol

Avanti Research - A Croda Brand 700156P, powder

Synonym(s):

5-cholestene-3α,20α-diol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C27H46 O2
CAS Number:
Molecular Weight:
402.65
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25
Pricing and availability is not currently available.

Assay

>99% (TLC)

form

powder

packaging

pkg of 1 × 10 mg (700156P-10mg)
pkg of 1 × 5 mg (700156P-5mg)

manufacturer/tradename

Avanti Research - A Croda Brand 700156P

shipped in

dry ice

storage temp.

−20°C

SMILES string

O[C@H]1CC[C@@]2([C@@H]3[C@H]([C@H]4[C@@]([C@H](CC4)[C@@](O)(CCCC(C)C)C)(CC3)C)CC=C2C1)C

InChI

1S/C27H46O2/c1-18(2)7-6-14-27(5,29)24-11-10-22-21-9-8-19-17-20(28)12-15-25(19,3)23(21)13-16-26(22,24)4/h8,18,20-24,28-29H,6-7,9-17H2,1-5H3/t20-,21-,22-,23-,24-,25-,26-,27-/m0/s1

InChI key

MCKLJFJEQRYRQT-APGJSSKUSA-N

Application

20α-hydroxycholesterol or 5-cholestene-3α,20α-diol has been used to study the role of oxysterols on mammalian immune function.[1]

Packaging

5 mL Amber Glass Screw Cap Vial (700156P-10mg)
5 mL Amber Glass Screw Cap Vial (700156P-5mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Michael E Abrams et al.
Nature microbiology, 5(7), 929-942 (2020-04-15)
Cholesterol 25-hydroxylase (CH25H) is an interferon-stimulated gene that converts cholesterol to the oxysterol 25-hydroxycholesterol (25HC). Circulating 25HC modulates essential immunological processes including antiviral immunity, inflammasome activation and antibody class switching; and dysregulation of CH25H may contribute to chronic inflammatory disease

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service