Skip to Content
Merck
All Photos(1)

Key Documents

700092P

Avanti

campesterol-d6

campesterol-d6, powder

Synonym(s):

26,27-hexadeuteriocampest-5-en-3β-ol; 111018

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C28H42D6O
CAS Number:
Molecular Weight:
406.72
MDL number:
UNSPSC Code:
41141804
NACRES:
NA.25

Assay

>99% (TLC)

form

powder

packaging

pkg of 1 × 1 mg (700092P-1mg)

manufacturer/tradename

Avanti Research - A Croda Brand 700092P

shipped in

dry ice

storage temp.

−20°C

General description

Campesterol-d6 is the deuterated form of campesterol. It is a plant sterol, which is an analog of cholesterol and a brassinosteroid (BR) precursor. The enzyme diminute/dwarf1 mediates the synthesis of campesterol from 24-methylenecholesterol. Campesterol possesses methyl group at C-24 position of the side chain in the cholesterol structure. It is not synthesized in mammanls and is diet derived

Application

Campesterol-d6 has been used as a deuterium-labeled internal standard in gas chromatography (GC)–triple quadrupole mass spectrometry and liquid chromatography-high resolution mass spectrometry (LC-HRMS).

Biochem/physiol Actions

Campesterol reduces the levels of chylomicron. The d6-campesterol along with d6-sitosterol and d6-cholesterol has been used in the transport blood-brain barrier studies in human brain endothelial hCMEC/D3 monolayer cells. Campesterol is a biomarker for tracking cholesterol synthesis and absorption. Campesterol like cholesterol is catabolized by intestinal microbes. It possess chemopreventive functionality prostate and breast tumors. Campesterol also has antiangiogenic property and may be useful in treating disorders related to angiogenesis.

Packaging

5 mL Amber Glass Screw Cap Vial (700092P-1mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S Matysik et al.
Clinical chemistry, 58(11), 1557-1564 (2012-09-22)
Cholesterol precursors and plant sterols have considerable potential as plasma biomarkers in several disorders of sterol metabolism and intestinal sterol absorption. Oxysterols are associated with atherogenesis, neurodegeneration, and inflammation. We developed a GC-MS method for the simultaneous analysis of these
Alan H B Wu et al.
Clinica chimica acta; international journal of clinical chemistry, 430, 43-47 (2014-01-08)
The analysis of blood for β-sitosterol and campesterol is the measures of cholesterol absorption while lathosterol is a measure of cholesterol synthesis. The biological variability of β-sitosterol, campesterol, and lathosterol was measured using liquid-chromatography tandem mass spectrometry from a cohort
Jung-Min Choi et al.
Phytotherapy research : PTR, 21(10), 954-959 (2007-07-03)
Campesterol, a plant sterol in nature, is known to have cholesterol lowering and anticarcinogenic effects. Since angiogenesis is essential for cancer, it was surmised that an antiangiogenic effect may be involved in the anticancer action of this compound. This study
Tim Vanmierlo et al.
Journal of lipid research, 53(4), 726-735 (2012-01-27)
Plant sterols such as sitosterol and campesterol are frequently administered as cholesterol-lowering supplements in food. Recently, it has been shown in mice that, in contrast to the structurally related cholesterol, circulating plant sterols can enter the brain. We questioned whether
Laura Calpe-Berdiel et al.
Atherosclerosis, 203(1), 18-31 (2008-08-12)
Plant sterols and stanols (phytosterols/phytostanols) are known to reduce serum low-density lipoprotein (LDL)-cholesterol level, and food products containing these plant compounds are widely used as a therapeutic dietary option to reduce plasma cholesterol and atherosclerotic risk. The cholesterol-lowering action of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service