610001
Avanti
Forceps 316SS
Avanti Research™ - A Croda Brand 610001
Synonym(s):
Forceps, Membrane Forceps 316SS
About This Item
Recommended Products
manufacturer/tradename
Avanti Research™ - A Croda Brand 610001
storage temp.
room temp
recommended
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service