Z742373
2mag mixBATH PC water bath
for use with MIXdrive 1/6/15/60 and MIXdrive 1 XS/1 eco, polycarbonate
Synonym(s):
polycarbonate water bath tank
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41103706
NACRES:
NB.72
Recommended Products
material
polycarbonate
packaging
pack of 1 ea
manufacturer/tradename
2mag 40850
parameter
35-72 °C temp. range
inside W × D × H
280 mm × 540 mm × 178 mm
outside W × D × H
300 mm × 560 mm × 198 mm
weight
6.2 kg
General description
Durable, robust bath, made of polycarbonate, easy to clean,
suitable for MIXdrive 1/6/15/60 and MIXdrive 1 XS/1 eco.
Water bath made of polycarbonate, durable up to +50 C,
robust, easy to clean, suitable for MIXdrive 1/6/15/60 as
well as MIXdrive 1 XS/1 eco, fitting for immersion thermostats
(heating/cooling), easy to clean, exact positioning of the
stirrer in the case of use the stirrer under the bath.
suitable for MIXdrive 1/6/15/60 and MIXdrive 1 XS/1 eco.
Water bath made of polycarbonate, durable up to +50 C,
robust, easy to clean, suitable for MIXdrive 1/6/15/60 as
well as MIXdrive 1 XS/1 eco, fitting for immersion thermostats
(heating/cooling), easy to clean, exact positioning of the
stirrer in the case of use the stirrer under the bath.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service