Merck
All Photos(4)
Key Documents
Z635545
Swiss Boy lab jacks
load ≤80 kg (with two plates, middle position), platform L × W 200 mm × 200 mm, Model 115
About This Item
UNSPSC Code:
41122800
NACRES:
NB.35
Pricing and availability is not currently available.
manufacturer/tradename
Grauer
parameter
≤80 kg load (with two plates, middle position)
76-280 mm vertical limit
platform L × W
200 mm × 200 mm
color
gold
General description
Jack can be raised with load in place.
These high quality Swiss made jacks have massive cast aluminum frames with a chemically resistant anodised finish. Swiss boy jacks are designed to carry heavy loads safely. Available either in standard size or with extended carrying plates for large pots or pans. Units carrying more that 90kg are also available with a motorized lift adjustment for enhanced convenience. The max load is for the middle position. Supplied with top and bottom plates.
Products with motorized jacks are available in Europe only.
Choose from one of the most recent versions:
Certificates of Analysis (COA)