Z530743
Space Saver vacuum manifold
positions, 8, L 420 mm
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
40161501
NACRES:
NB.79
Recommended Products
feature
positions 8
L
420 mm
Looking for similar products? Visit Product Comparison Guide
General description
Designed for laboratories where space is at a premium.
Features:
Features:
- Takes up 35% less space than the traditional 5-position manifold
- Compact size with ample space for lattice clamps
- High-vacuum PTFE valves
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service