Key Documents
Z507873
Three-neck round-bottom flasks
capacity 25 mL
About This Item
UNSPSC Code:
41121804
NACRES:
NB.43
Pricing and availability is not currently available.
material
glass
round bottom flask
three-neck
sterility
non-sterile
feature
closure type Holed cap
packaging
pack of 1
manufacturer/tradename
Ace Glass 9465-34
capacity
25 mL
General description
<ST/>14/10 threaded joints. Supplied with holed caps, PTFE-faced silicone septa, and FETFE® O-rings.
Legal Information
FETFE is a registered trademark of Ace Glass, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)