Key Documents
Z301140
Quickfit® expansion adapter
top female joint: ST/NS 29/32, bottom male joint: ST/NS 24/29
Synonym(s):
XA43
About This Item
UNSPSC Code:
41121815
NACRES:
NB.35
Pricing and availability is not currently available.
Joint
female joint: ST/NS top29/32
male joint: ST/NS bottom24/29
manufacturer/tradename
Quickfit XA 43
availability
not available in USA and Canada
not available in USA and Canada
General description
These adapters connect items with larger cone to a piece of glassware with a smaller size socket.
Legal Information
Quickfit is a registered trademark of SciLabware Limited
Choose from one of the most recent versions:
Certificates of Analysis (COA)