Z263478
Wheaton straight-mouth bottles with cap, clear
clear soda-lime glass bottle, capacity 500 mL, white polypropylene cap, (PTFE faced LDPE liner), case of 12 ea
Synonym(s):
Wheaton bottles, bottles clear, clear bottles
About This Item
Recommended Products
material
(PTFE faced LDPE liner)
clear soda-lime glass bottle
round bottle
white polypropylene cap
Joint
Joints threaded neck (89-400)
feature
graduations: no
with cap
packaging
case of 12 ea
manufacturer/tradename
WHEATON® W216911
capacity
500 mL
diam. × H
91 mm × 95 mm
Looking for similar products? Visit Product Comparison Guide
General description
Application
Features and Benefits
Legal Information
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service