Key Documents
Z220140
Sigma-Aldrich
Filter funnel with drip-tip and two side-arms each equipped with a glass stopcock
capacity 100 mL, joint: ST/NS 14/20 M
About This Item
UNSPSC Code:
41104900
NACRES:
NB.43
Pricing and availability is not currently available.
material
borosilicate glass
label
Y
Joint
joint ST/NS14/20 F
joint: ST/NS14/20 M
manufacturer/tradename
Aldrich
capacity
100 mL
pore size
25-50 μm porosity
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Visit the Document Library
Articles
Versatile system for chemical reactions, extractions, and transfers of solids and liquids in laboratories.
Versatile system for chemical reactions, extractions, and transfers of solids and liquids in laboratories.
Versatile system for chemical reactions, extractions, and transfers of solids and liquids in laboratories.
Versatile system for chemical reactions, extractions, and transfers of solids and liquids in laboratories.