Key Documents
Z193194
Metric mini ruler
stainless steel, thickness × W × L 1 mm × 1 cm × 100 mm
About This Item
UNSPSC Code:
41111700
NACRES:
NB.79
Pricing and availability is not currently available.
material
stainless steel
thickness × W × L
1 mm × 1 cm × 100 mm
General description
Pocket size ruler.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Questions
1–2 of 2 Questions  
  1. Is this ruler sterilizable/autoclavable?
    1 answer
    1. This item is autoclavable. The long term stability of the lettering following multiple sterilization events has not been evaluated.
      Helpful?
  2. Hello, does this ruler come with a calibration certificate?
    1 answer
    1. There is no calibration certificate for this rule.
      Helpful?