Z188778
Wheaton sample vials
amber, volume 2 mL, cap size 8 - 425, diam. × H 12 mm × 36 mm
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41122001
NACRES:
NB.11
Recommended Products
material
amber borosilicate glass vial
feature
closure type screw top vial
packaging
pkg of 200 ea
cap size
8 - 425
diam. × H
12 mm × 36 mm
volume
2 mL
color
amber
fitting
thread for 8-425
Looking for similar products? Visit Product Comparison Guide
General description
Vials are packaged in corrugated trays with partitions then shrink-wrapped to reduce particulate contamination.
Autoclavable borosilicate USP Type 1 glass. Order caps separately.
Autoclavable borosilicate USP Type 1 glass. Order caps separately.
related product
Product No.
Description
Pricing
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service