Key Documents
Z170259
Lightnin® 316 stainless steel impeller
3 Blade turbine, Model A-310, diam. 2 1/2 in. , overall
About This Item
UNSPSC Code:
41100000
NACRES:
NB.72
Pricing and availability is not currently available.
manufacturer/tradename
Model A-310
Lightnin 808955
bore size
5/16 in.
diam.
2 1/2 in. , overall
General description
High efficiency design develops 50% more mixing action and true axial flow.
Legal Information
Lightnin is a registered trademark of Lightnin, a unit of General Signal
Choose from one of the most recent versions:
Certificates of Analysis (COA)