Key Documents
Z150754
Water aspirator pump
Chapman swirling
About This Item
UNSPSC Code:
41105100
NACRES:
NB.79
L
137 mm
Related Categories
General description
For greater air-entrainment capacity.
Operates efficiently on both high and low water pressure. Nickel-plated 3/8 in. NPTM water inlet. Ball-type, positive-acting valve in suction tube prevents backflow should pressure be reduced. Max. vacuum is 100 mm Hg (abs).
Choose from one of the most recent versions:
Certificates of Analysis (COA)